Skip to Content
Merck
All Photos(2)

Key Documents

SMB00344

Sigma-Aldrich

Pectolinarin

≥98% (HPLC)

Synonym(s):

Pectolinarin, 7-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-5-hydroxy-6-methoxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, Pectolinaroside, 5,7-Dihydroxy 4′,6-dimethoxyflavone 7-rutinoside, Pectolinarigenin 7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H34O15
CAS Number:
Molecular Weight:
622.57
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC1=C(C(C=C(C2=CC=C(C=C2)OC)O3)=O)C3=CC(O[C@H]4[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO[C@H]5[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O5)O4)=C1OC

InChI

1S/C30H36O14/c1-12-22(32)26(36)28(38)30(42-12)41-11-14-8-19(24(34)27(37)23(14)33)44-20-10-18-21(25(35)29(20)40-3)16(31)9-17(43-18)13-4-6-15(39-2)7-5-13/h4-7,9-10,12,14,19,22-24,26-28,30,32-38H,8,11H2,1-3H3/t12-,14+,19+,22-,23+,24-,26+,27-,28+,30+/m0/s1

InChI key

PRVAKFUXKKVCCY-YLHHYQKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pectolinarinis a glycosylated flavonoid that is generally found in several Cirsium spp. and was first extracted from Linaria vulgaris. The chemical structure has a rutinoside conjugate of pectolinarigenin at the 7-O position.

Biochem/physiol Actions

Pectolinarinexerts anti-inflammatory, antioxidant, antidiabetic, and antitumor activities. It is known to be a potent agent for the treatment of neurodegenerative disorders. It also displays antiviral, and antiparasitic effects and can inhibit bacterial biofilm formation. Pectolinarin relieves lipopolysaccharide-induced liver and kidney injuries.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification Process and In Vitro and In Vivo Bioactivity Evaluation of Pectolinarin and Linarin from Cirsium japonicum
Ye Yana, et al.,
Molecules (Basel), 27 (24), 8695-8695 (2022)
P-EP017. Biological potential and neuroprotective activity of pectolinarin in the management of neurodegenerative disorders: Phytotherapeutic importance through data analysis of various scientific works
Patel Dinesh Kumar and Patel Kanika
Clinical Neurophysiology : Official Journal of the International Federation of Clinical Neurophysiology, 132(8), e83-e84 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service