Skip to Content
Merck
All Photos(1)

Key Documents

SMB00246

Sigma-Aldrich

Calceolarioside A

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26O11
CAS Number:
Molecular Weight:
478.45
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OCCc2ccc(O)c(O)c2)[C@H](O)[C@@H](O)[C@@H]1OC(=O)\C=C\c3ccc(O)c(O)c3

InChI

1S/C23H26O11/c24-11-18-22(34-19(29)6-3-12-1-4-14(25)16(27)9-12)20(30)21(31)23(33-18)32-8-7-13-2-5-15(26)17(28)10-13/h1-6,9-10,18,20-28,30-31H,7-8,11H2/b6-3+/t18-,20-,21-,22-,23-/m1/s1

InChI key

UHIGZYLCYRQESL-VJWFJHQPSA-N

General description

Natural product derived from plant source.

Application

Calceolarioside A has been used in flow cytometric analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Damtoft et al.
Phytochemistry, 37(2), 441-443 (1994-09-01)
From Chirita sinensis (Gesneriaceae) was isolated six phenylethanoid glucosides, namely the previously known positional isomers plantainoside A, calceolarioside A and calceolarioside B together with three new compounds named chiritoside A, B and C. It was proved by analysis of the
Avijit Poddar et al.
Planta medica, 74(5), 503-508 (2008-06-11)
Bioactivity-guided fractionation has led to the successful isolation of calceolarioside A ( 1) from the methanolic extract of night jasmine leaves. The in vitro antileishmanial activity of compound 1 was determined (IC (50) = 20 microg/mL). Its IN VIVO efficacy
A Capasso et al.
Planta medica, 59(4), 337-339 (1993-08-01)
The effect of calceolarioside A, a phenylpropanoid glycoside (PhG), isolated from Calceolaria hypericina, was studied on rabbit platelets in vitro. Calceolarioside A induced a dose-related aggregant effect on rabbit platelets. Indomethacin did not modify the calceolarioside A-induced aggregant effect. Furthermore
José G Sena Filho et al.
Journal of natural products, 72(7), 1344-1347 (2009-06-03)
Many references to the use of Lantana spp. can be found in the ethnopharmacological literature from locations around the globe. This study was focused on examining constituents from the polar extracts of Lantana radula Sw. and Lantana canescens Kunth, for
Targeting Malassezia species for novel synthetic and natural antidandruff agents.
Letizia A, et al.
Current Medicinal Chemistry, 24(22), 2392-2412 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service