Skip to Content
Merck
All Photos(1)

Key Documents

68783

Sigma-Aldrich

Licochalcone A

≥96.0% (HPLC)

Synonym(s):

(E)-3-[5-(1,1-Dimethyl-2-propenyl)-4-hydroxy-2-methoxyphenyl]-1-(4-hydroxyphenyl)-2-propen-1-one, 4′,4-Dihydroxy-3-α,α-dimethylallyl-6-methoxychalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
Beilstein:
4534154
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥96.0% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

COc1cc(O)c(cc1\C=C\C(=O)c2ccc(O)cc2)C(C)(C)C=C

InChI

1S/C21H22O4/c1-5-21(2,3)17-12-15(20(25-4)13-19(17)24)8-11-18(23)14-6-9-16(22)10-7-14/h5-13,22,24H,1H2,2-4H3/b11-8+

InChI key

KAZSKMJFUPEHHW-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Licochalcone A is a potent membrane-active agent that transforms normal erythrocytes into echinocytes. A novel flavanoid which has demonstrated significant antitumor activity.

Biochem/physiol Actions

Inhibits vegetative growth of spore-forming bacteria, B. subtilis and other food-contaminating microorganisms.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryo-Ichi Tsukiyama et al.
Antimicrobial agents and chemotherapy, 46(5), 1226-1230 (2002-04-18)
Licochalcone A was isolated from the roots of licorice, Glycyrrhiza inflata, which has various uses in the food and pharmaceutical industries; isolation was followed by extraction with ethanol and column chromatography with silica gel. In this study, the activities of
Xiu-ying Xiao et al.
Cancer letters, 302(1), 69-75 (2011-01-11)
The aim of this study was to determine the anticancer effects of seven licorice compounds in MKN-28, AGS, and MKN-45 gastric cancer cells and human gastric epithelium immortalized cells. We also explored the mechanism of action of licochalcone A (LCA)
Ewelina Szliszka et al.
International journal of molecular sciences, 11(1), 1-13 (2010-02-18)
Chalcones exhibit chemopreventive and antitumor effects. Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a naturally occurring anticancer agent that induces apoptosis in cancer cells and is not toxic to normal cells. We examined the cytotoxic and apoptotic effect of five
M Udompataikul et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 25(6), 660-665 (2010-09-16)
Although moisturizer usage has been considered a mainstay of treatment for atopic dermatitis (AD) patients, few clinical studies have been investigated. Recently, moisturizers containing non-steroidal anti-inflammatory agents, such as licochalcone A (LA) and vitamin B(12) are of emerging interest. To
Sandra Monti et al.
Physical chemistry chemical physics : PCCP, 10(44), 6597-6606 (2008-11-08)
In the present contribution we address the study of the interaction of a flavonoid-derivative licochalcone A (LA) with human serum albumin (HSA). The application of circular dichroism, UV-Vis absorption, fluorescence and laser flash photolysis combined with molecular mechanics, molecular dynamics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service