Skip to Content
Merck
All Photos(1)

Key Documents

SHH0023

Sigma-Aldrich

PolyFreeze Tissue Freezing Medium

green

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

color

green

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

PolyFreeze Tissue Freezing Medium does not result in autofluorescence. It is easy to cleanse at the time of tissue fixation, when washed before staining. After washing the medium, there will be no trace of the support matrix to hinder with staining or immunohistochemistry (IHC) reactions.

Application

A support matrix or form of embedding medium for frozen sectioning. This medium freezes quickly supporting the tissue for sectioning at 3 μ and up with no cracking of the matrix at temperatures from -8 °C to -25 °C. Snap freeze using PolyFreeze with isopentane and liquid nitrogen or dry ice (slush/slurry or bunker). Store specimens frozen in PolyFreeze in liquid nitrogen canisters or in airtight containers in a -80 °C freezer.

Features and Benefits

  • Colored PolyFreeze (blue, green, yellow) makes differentiating multiple specimens easy during sectioning and staining
  • Experience less curling, less ice artifacts, faster freezing with PolyFreeze

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service