Skip to Content
Merck
All Photos(3)

Key Documents

277258

Sigma-Aldrich

2-Methylbutane

anhydrous, ≥99%

Synonym(s):

Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Beilstein:
1730723
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
30 °C (lit.)
vapor pressure:
11.17 psi ( 20 °C)

grade

anhydrous

Quality Level

vapor density

2.6 (vs air)

vapor pressure

11.17 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

788 °F

expl. lim.

8.3 %

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.354 (lit.)

bp

30 °C (lit.)

mp

-160 °C

density

0.62 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylbutane undergoes catalytic dehydrogenation in the presence of chromia-alumina catalyst to form isoamylenes, which can undergo further dehydrogenation to form isoprene. It may also be used as a solvent in the preparation of trans-Bis(triethylphosphine) (hydroxy carbonyl) (phenyl) platinum(II), a metallacarboxylic acid.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiming Huang et al.
Transplantation, 89(6), 677-685 (2010-01-30)
The role of bone marrow (BM)-derived cells in pancreatic beta-cell regeneration remains unresolved. We examined whether BM-derived cells are recruited to the site of moderate pancreatic injury and contribute to beta-cell regeneration. Low-dose streptozotocin (STZ) treatment was used to induce
Power efficient synthesis of isoprene via two-stage dehydrogenation of isopentane
Karimov OK &amp; Daminev RR
World Applied Sciences Journal , 24(3), 320-325 (2013)
Johnson AW.
Invitation to Organic Chemistry., 338-338 (1999)
Synthesis, x-ray structural analysis, and thermal decomposition of the platinum (II) carboxylic acid (hydroxycarbonyl) trans-Pt (CO2H)(C6H5)(PEt3) 2. Formation of a diplatinum (II) complex containing carbon dioxide
Bennett MA, et al
Journal of the American Chemical Society, 110(21), 7098-7105 (1988)
Rotational Isomerism and Thermodynamic Functions of 2-Methylbutane and 2, 3-Dimethylbutane. Vapor Heat Capacity and Heat of Vaporization of 2-Methylbutane.
Scott DW, et al.
Journal of the American Chemical Society, 73(4), 1707-1712 (1951)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service