Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700340

Sigma-Aldrich

Monoclonal Anti-IFNG antibody produced in mouse

clone G-23, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-IFN-γ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

G-23, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IFNG(3458)

General description

The antibody G-23 reacts with IFN-gamma, a 20-25 kDa cytokine produced by activated Th1 cells and NK cells.

Immunogen

Recombinant human IFN-gamma (aa 22-166 representing mature IFN-gamma)

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 2-4 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Laurin et al.
Journal of biomedicine & biotechnology, 2010, 234540-234540 (2010-07-10)
Adoptive transfer of specific cytotoxic T lymphocytes (CTL) and Cytokine Induced Killer Cells (CIK) following genetic engineering of T-cell receptor zeta hold promising perspective in immunotherapy. In the present work we focused on the mechanisms of anti-tumor action of effectors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service