Skip to Content
Merck
All Photos(2)

Documents

SAB4501933

Sigma-Aldrich

Anti-MUC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Breast carcinoma-associated antigen DF3, CD227, Carcinoma-associated mucin, EMA, Episialin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 122 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC1(4582)

General description

Anti-MUC1 Antibody detects endogenous levels of total MUC1 protein.
MUC1 (Mucin 1) is O-glycosylated, carbohydrate rich, transmembrane glycoprotein. It has approximately 13 members in the family. It is localized on the apical surface of mucosal epithelia in the lung, eye, breast and stomach. It is overexpressed in several epithelial cancers, including pancreatic ductal adenocarcinoma. It is a heterodimer consisting of N-terminal (MUC1-N) and C-terminal (MUC1-C) subunits. N-terminal (MUC1-N) end contains tandem repeats (VNTR) which forms the mucin component.

Immunogen

The antiserum was produced against synthesized peptide derived from human MUC1.

Immunogen Range: 1191-1240

Application

Anti-MUC1, C-Terminal antibody produced in rabbit is suitable for immunohistochemistry and indirect immunofluorescence.

Biochem/physiol Actions

MUC1 (Mucin 1) plays a major role in the formation of epithelium lining of the gastrointestinal tract. It lubricates the gastrointestinal tract, to protect the epithelia from oesophagus to the colon. It also lubricates the epithelia of some organs such as pancreas and gall bladder. It is directly involved in several signaling pathways as a modulator that affects oncogenesis, motility and metastasis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P Corfield et al.
Frontiers in bioscience : a journal and virtual library, 6, D1321-D1357 (2001-10-02)
Mucins form part of the dynamic, interactive mucosal defensive system active at the mucosal surface of the gastrointestinal tract. They are carbohydrate rich glycoproteins with unique molecular structure and chemical properties. The family of mucin (MUC) genes has 13 members
C J Reid et al.
Gut, 42(2), 220-226 (1998-04-16)
Mucin glycoproteins play a key role in the normal function of the epithelium lining the gastrointestinal tract. The expression of mucin genes, MUC 3, 4, 5AC, 5B, 6, 7, and 8 in human fetal tissues was examined to establish the
M Sahraei et al.
Oncogene, 31(47), 4935-4945 (2012-01-24)
Pancreatic ductal adenocarcinoma (PDA) has one of the worst prognoses of all cancers. Mucin 1 (MUC1), a transmembrane mucin glycoprotein, is a key modulator of several signaling pathways that affect oncogenesis, motility and metastasis. Its expression is known to be
Lili Zhang et al.
The Journal of pathology, 234(1), 60-73 (2014-05-20)
Cigarette smoke increases the risk of lung cancer by 20-fold and accounts for 87% of lung cancer deaths. In the normal airway, heavily O-glycosylated mucin-1 (MUC1) and adherens junctions (AJs) establish a structural barrier that protects the airway from infectious
Solange Tréhoux et al.
Biochemical and biophysical research communications, 456(3), 757-762 (2014-12-17)
MUC1 is an oncogenic mucin overexpressed in several epithelial cancers, including pancreatic ductal adenocarcinoma, and is considered as a potent target for cancer therapy. To this aim, we undertook to study MUC1 biological effects on pancreatic cancer cells and identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service