Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501452

Sigma-Aldrich

Anti-GRP78 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

78 kDa glucose-regulated protein, BiP, GRP-78, Heat shock 70 kDa protein 5, Immunoglobulin heavy chain-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 72 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA5(3309)

General description

Glucose/regulated protein 78 (GRP78) is a stress-associated protein. It is mapped to human chromosome 9q33.3. It is also known as HSPA5 (heat shock 70kDa protein 5) and immunoglobulin heavy chain binding protein (BiP). It is present in nucleated cells like thyroid, lung, smooth muscle, liver and several cells of the immune system.
The molecular weight of GRP78 is 78kDa.

Immunogen

The antiserum was produced against synthesized peptide derived from human GRP78.

Immunogen Range: 605-654

Biochem/physiol Actions

GRP78 levels are elevated during various human cancers and tumors.
Glucose/regulated protein 78 (GRP78) participates in stress-associated diseases. It also participates in protein biogenesis (protein transport into the endoplasmic reticulum (ER), protein folding and assembly and ER associated protein degradation), signal transduction and calcium homeostasis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HSPA5 (heat shock 70kDa protein 5 (glucoseregulated protein, 78kDa)).
Zimmermann R and Dudek J
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2010)
Overexpression of the glucose-regulated stress gene GRP78 in malignant but not benign human breast lesions
Fernandez P M, et al.
Breast Cancer Research and Treatment, 59(1), 15-26 (2000)
Role of the unfolded protein response regulator GRP78/BiP in development, cancer, and neurological disorders
Wang M, et al.
Antioxidants & Redox Signaling, 11(9), 2307-2316 (2009)
Polymorphisms of glucose-regulated protein 78 and clinical relevance of neuroblastoma: Risk and prognosis.
Zhang J, et al.
Journal of Cancer Research and Therapeutics, 12(3), 1178-1178 (2016)
Genetic polymorphisms and haplotype structures of HSPA5 gene in the Han population of Southern China.
Zhu X, et al.
Tissue Antigens, 74(5), 420-423 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service