Skip to Content
Merck
All Photos(1)

Documents

SAB1305552

Sigma-Aldrich

MONOCLONAL ANTI-LC3 (APG8) antibody produced in mouse

clone 166AT1234, IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, MAP1 light chain 3-like protein 1, MAP1LC3A, Microtubule-associated proteins 1A/1B light chain 3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

166AT1234, monoclonal

form

buffered aqueous solution

mol wt

14272 Da

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: 1:25
immunohistochemistry: 1:50-1:100
western blot: 1:1000

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoming Shu et al.
Molecular medicine reports, 16(2), 1180-1188 (2017-06-07)
Peripheral blood T lymphocytopenia has previously been identified in polymyositis/dermatomyositis (PM/DM) patients. Therefore, the present study aimed to examine the potential role of autophagy in peripheral blood T cell survival in PM/DM patients. Transmission electron microscopy was used to detect
Hong Ding et al.
Journal of asthma and allergy, 17, 717-731 (2024-08-06)
Accumulating evidence indicates that oxidative stress and inflammation are the pathological basis of allergic diseases. Inhibition of NOD-like receptor family pyrin domain-containing 3 (NLRP3) inflammasome could ameliorate allergic rhinitis (AR). Here, we explored the effects and mechanisms that underlie NLRP3
Maomao Sun et al.
Frontiers in immunology, 12, 685523-685523 (2021-08-03)
Recent studies have shown that autophagy upregulation can attenuate sepsis-induced acute kidney injury (SAKI). The tumor suppressor p53 has emerged as an autophagy regulator in various forms of acute kidney injury (AKI). Our previous studies showed that p53 acetylation exacerbated
Matteo Bordoni et al.
Cells, 11(8) (2022-04-24)
Mitochondria alterations are present in tissues derived from patients and animal models, but no data are available for peripheral blood mononuclear cells (PBMCs) of ALS patients. This work aims to investigate mitophagy in PBMCs of sporadic (sALS) patients and how
Rongrong Hua et al.
Journal of cellular biochemistry, 120(9), 15915-15923 (2019-05-14)
The sequential reactivation of mechanistic target of rapamycin (mTOR) inhibited autophagic flux in neurons exposed to oxygen-glucose deprivation/reperfusion (OGD/R), which was characterized by reduction of autophagosome formation and restriction of autolysosome degradation. However, its detailed molecular mechanism was still unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service