Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301607

Sigma-Aldrich

Anti-CD63

affinity isolated antibody

Synonym(s):

LAMP-3, ME491, MLA1, OMA81H, TSPAN30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:25-1:100
western blot: 1:500-1:2000

accession no.

NP_001244318

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD63(967)

General description

Cluster of differentiation (CD63) belongs to the tetraspanin family, with four transmembrane domains. It is localised in late endosomes and lysosomes. The gene is located on human chromosome 12q13.2. CD63 is a membrane bound glycoprotein.

Immunogen

Synthetic peptide of human CD63 molecule

Biochem/physiol Actions

Cluster of differentiation (CD63) acts as a platelet activation marker. It plays an important role in regulating cell proliferation, adhesion, motility and differentiation. CD63 acts as a prognostic marker for lung adenocarcinoma. Deficiency of CD63 is associated with Hermansky−Pudlak syndrome (HPS).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongdong Tian et al.
International journal of molecular medicine, 50(1) (2022-05-28)
Chronic obstructive pulmonary disease (COPD) is one of the major causes of death worldwide today, and its related morbidity has been predicted to show an increase in subsequent years. Recent studies have shown that Danshen, a Chinese herbal medicine, is
Hermansky-Pudlak syndrome: a disease of protein trafficking and organelle function
Wei ML
Pigment Cell Research / Sponsored by the European Society for Pigment Cell Research and the International Pigment Cell Society, 19(1), 19-42 (2006)
Trafficking and function of the tetraspanin CD63
Pols MS and Klumperman J
Experimental Cell Research, 315(9), 1584-1592 (2009)
Comprehensive whole genome array CGH profiling of mantle cell lymphoma model genomes
De Leeuw RJ, et al.
Human Molecular Genetics, 13(17), 1827-1837 (2004)
Course of platelet activation markers after ischemic stroke
Marquardt L, et al.
Stroke, 33(11), 2570-2574 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service