Skip to Content
Merck
All Photos(1)

Documents

C9993

Sigma-Aldrich

Anti-CD9 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BA2, Anti-BTCC-1, Anti-DRAP-27, Anti-GIG2, Anti-Growth-inhibiting gene 2 protein, Anti-Leukocyte antigen MIC3, Anti-MRP-1, Anti-Motility related protein, Anti-TSPAN29, Anti-p24 antigen

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~25 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:250
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD9(928)
mouse ... Cd9(12527)
rat ... Cd9(24936)

General description

CD9 is a glioma stem cell (GSC)-enriched protein, which belongs to tetraspanin family. It is a cell motility related molecule. The gene is located on human chromosome 12p13.31. CD9 is expressed on pre-B cells, activated T cells and Schwann cells.

Immunogen

synthetic peptide corresponding to residues 36-50 of human CD9.

Application

Anti-CD9 antibody produced in rabbit has been used in
  • immunostaining
  • identification of hypoxia-induced mesenchymal stem cells (MSC-MVs)
Anti-CD9 antibody produced in rabbit is suitable for immunohistochemistry at a working dilution of 1:100-1:250 and western blotting at a working dilution of 1:500-1:1000. It was used as a primary antibody for the immunolocalization of adhesion protein CD9 during a study to investigate the impact of adhesion proteins on skeletal myoblast differentiation.

Biochem/physiol Actions

CD9 plays a major role in sperm-egg fusion in fertilization. It enhances cancer invasion and tumor growth in cervical cancer, gastric cancer and glioblastoma (GBM). CD9 is involved in cell invasion, apoptosis and resistance to chemotherapy.

Target description

The CD9 protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It can modulate cell adhesion and migration. It also triggers platelet activation and aggregation. In addition, the protein appears to promote muscle cell fusion and support myotube maintenance.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karolina Archacka et al.
Stem cells and development, 23(20), 2455-2468 (2014-06-19)
Pluripotent stem cells are a potential source of various cell types for use in regenerative medicine. Despite accumulating knowledge, there is currently no efficient and reproducible protocol that does not require genetic manipulation for generation of myogenic cells from pluripotent
CD9 suppresses human extravillous trophoblast invasion
Matsumoto H, et al.
Placenta, 47(1), 105-112 (2016)
Competence of in vitro cultured mouse embryonic stem cells for myogenic differentiation and fusion with myoblasts
Archacka K, et al.
Stem Cells and Development, 23(20), 2455-2468 (2014)
Retinoic acid represses a cassette of candidate pluripotency chromosome 12p genes during induced loss of human embryonal carcinoma tumorigenicity
Giuliano CJ, et al.
Biochimica et Biophysica Acta (BBA)-Gene Structure and Expression, 1731(1), 48-56 (2005)
Surface phosphatidylserine is responsible for the internalization on microvesicles derived from hypoxia-induced human bone marrow mesenchymal stem cells into human endothelial cells
Wei X, et al.
PLoS ONE, 11(1), e0147360-e0147360 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service