Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410565

Sigma-Aldrich

Anti-CYP4V2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BCD, CYP4AH1, FLJ18432, MGC43534

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP4V2(285440)

General description

Cytochrome P450 4V2 (CYP4V2) belongs to human cytochrome P450 enzyme family and encodes a protein of 55 kDa. CYP4V2 gene is mapped to human chromosome 4q35.1−q35.2. CYP4V2 is majorly expressed in the epithelial cells of the retina and cornea. In retinal pigment epithelia (RPE) cell lines, the enzyme is localized to endoplasmic reticulum.
This gene encodes a member of the cytochrome P450 hemethiolate protein superfamily which are involved in oxidizing various substrates in the metabolic pathway. It is implicated in the metabolism of fatty acid precursors into n-3 polyunsaturated fatty acids. Mutations in this gene result in Bietti crystalline corneoretinal dystrophy. (provided by RefSeq)

Immunogen

CYP4V2 (NP_997235.2, 1 a.a. ~ 525 a.a) full-length human protein.

Sequence
MAGLWLGLVWQKLLLWGAASALSLAGASLVLSLLQRVASYARKWQQMRPIPTVARAYPLVGHALLMKPDGREFFQQIIEYTEEYRHMPLLKLWVGPVPMVALYNAENVEVILTSSKQIDKSSMYKFLEPWLGLGLLTSTGNKWRSRRKMLTPTFHFTILEDFLDIMNEQANILVKKLEKHINQEAFNCFFYITLCALDIICETAMGKNIGAQSNDDSEYVRAVYRMSEMIFRRIKMPWLWLDLWYLMFKEGWEHKKSLKILHTFTNSVIAERANEMNANEDCRGDGRGSAPSKNKRRAFLDLLLSVTDDEGNRLSHEDIREEVDTFMFEGHDTTAAAINWSLYLLGSNPEVQKKVDHELDDVFGKSDRPATVEDLKKLRYLECVIKETLRLFPSVPLFARSVSEDCEVAGYRVLKGTEAVIIPYALHRDPRYFPNPEEFQPERFFPENAQGRHPYAYVPFSAGPRNCIGQKFAVMEEKTILSCILRHFWIESNQKREELGLEGQLILRPSNGIWIKLKRRNADER

Biochem/physiol Actions

Cytochrome P450 4V2 (CYP4V2) is involved in the fatty acid ω-hydroxylation. It alters lipid homeostasis when expressed in human liver cancer HepG2 cells. It coordinates with enzymes involved in mitochondrial and peroxisomal α/β-oxidation. Functional CYP4V2 gene mutant is implicated in ocular disease called the Bietti crystalline dystrophy. It is also expressed in macrophages and is regulated by nuclear receptor peroxisome proliferator activated receptor γ (PPARγ).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and characterization of CYP4V2 as a fatty acid omega-hydroxylase
Nakano M, et al.
Drug Metabolism and Disposition, 37(11), 2119-2122 (2009)
Expression of CYP4V2 in human THP1 macrophages and its transcriptional regulation by peroxisome proliferator-activated receptor gamma
Yi M, et al.
Toxicology and Applied Pharmacology, 330(5), 100-106 (2017)
CYP4V2 in Bietti's crystalline dystrophy: ocular localization, metabolism of omega-3-polyunsaturated fatty acids, and functional deficit of the p. H331P variant
Nakano M, et al.
Molecular Pharmacology, 82(4), 679-686 (2012)
Jiang-Hui Wang et al.
Scientific reports, 12(1), 9525-9525 (2022-06-11)
Bietti crystalline dystrophy (BCD) is an inherited retinal disease (IRD) caused by mutations in the CYP4V2 gene. It is a relatively common cause of IRD in east Asia. A number of features of this disease make it highly amenable to
Erin L Heinzen et al.
American journal of human genetics, 86(5), 707-718 (2010-04-20)
Deletions at 16p13.11 are associated with schizophrenia, mental retardation, and most recently idiopathic generalized epilepsy. To evaluate the role of 16p13.11 deletions, as well as other structural variation, in epilepsy disorders, we used genome-wide screens to identify copy number variation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service