Skip to Content
Merck
All Photos(3)

Key Documents

SAB1408000

Sigma-Aldrich

Anti-GRHL2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BOM, DFNA28, FLJ11172, FLJ13782, MGC149294, MGC149295, TFCP2L3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~71.1 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRHL2(79977)

General description

Grainyhead like transcription factor 2 (GRHL2) encodes a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28). Grainyhead like transcription factor 2 (GRHL2) is an epithelial regulatory factor. GRHL2 is homologous to Drosophila grainyhead. GRHL2 gene is located on human chromosome 8q22.3.

Immunogen

GRHL2 (NP_079191.1, 1 a.a. ~ 625 a.a) full-length human protein.

Sequence
MSQESDNNKRLVALVPMPSDPPFNTRRAYTSEDEAWKSYLENPLTAATKAMMSINGDEDSAAALGLLYDYYKVPRDKRLLSVSKASDSQEDQEKRNCLGTSEAQSNLSGGENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSTPDSTYSESFKDAATEKFRSASVGAEEYMYDQTSSGTFQYTLEATKSLRQKQGEGPMTYLNKGQFYAITLSETGDNKCFRHPISKVRSVVMVVFSEDKNRDEQLKYWKYWHSRQHTAKQRVLDIADYKESFNTIGNIEEIAYNAVSFTWDVNEEAKIFITVNCLSTDFSSQKGVKGLPLMIQIDTYSYNNRSNKPIHRAYCQIKVFCDKGAERKIRDEERKQNRKKGKGQASQTQCNSSSDGKLAAIPLQKKSDITYFKTMPDLHSQPVLFIPDVHFANLQRTGQVYYNTDDEREGGSVLVKRMFRPMEEEFGPVPSKQMKEEGTKRVLLYVRKETDDVFDALMLKSPTVKGLMEAISEKYGLPVEKIAKLYKKSKKGILVNMDDNIIEHYSNEDTFILNMESMVEGFKVTLMEI

Application

Anti-GRHL2 antibody produced in mouse has been used in:
  • Immunofluorescence
  • Western blotting
  • Co-immunoprecipitation
  • Immunohistochemistry

Biochem/physiol Actions

Grainyhead like transcription factor 2 (GRHL2) inhibits epithelial-to-mesenchymal transition, which is necessary for cell migration. It is involved in neural tube closure, epithelial morphogenesis and barrier formation during embryogenesis. It contributes to carcinogenesis. GRHL2 regulates epithelial plasticity of pancreatic ductal adenocarcinoma (PDAC) progression.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grhl2 regulation of SPINT1 expression controls salivary gland development
Matsushita T, et al.
Biochemical and Biophysical Research Communications, 137(22) (2018)
Bridging cancer biology with the clinic: relative expression of a GRHL2-mediated gene-set pair predicts breast cancer metastasis
Yang X, et al.
PLoS ONE, 8(2), e56195-e56195 (2013)
Grainyhead-like 2 in development and cancer
Ma L, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(5), 1010428317698375-1010428317698375 (2017)
Novel androgen receptor co-regulator GRHL2 exerts both oncogenic and anti-metastatic functions in prostate cancer
Paltoglou S, et al.
Cancer Research, 137(22) (2017)
Grainyhead-like 2 (GRHL 2) regulates epithelial plasticity in pancreatic cancer progression
Nishino H, et al.
Cancer Medicine, 6(11) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service