Skip to Content
Merck
All Photos(8)

Key Documents

HPA006420

Sigma-Aldrich

Anti-GRHL1 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Grainyhead-like protein 1 homolog antibody produced in rabbit, Anti-Mammalian grainyhead antibody produced in rabbit, Anti-NH32 antibody produced in rabbit, Anti-Transcription factor CP2-like 2 antibody produced in rabbit, Anti-Transcription factor LBP-32 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VTVSIATMPTHSIKTETQPHGFAVGIPPAVYHPEPTERVVVFDRNLNTDQFSSGAQAPNAQRRTPDSTFSETFKEGVQEVFFPSDLSLRMPGMNSEDYVFDSVSGNNFEYTLEASKSLRQKPGDSTMTYLNK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRHL1(29841)

Looking for similar products? Visit Product Comparison Guide

General description

GRHL1 (grainyhead-like transcription factor 1) is a transcription factor, which is a member of the GRHL family, which also includes GRHL2 and GRHL3. This gene has a high level of expression in developing mouse skin. This gene is localized to human chromosome 2p25.1.

Immunogen

Grainyhead-like protein 1 homolog recombinant protein epitope signature tag (PrEST)

Application

Anti-GRHL1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GRHL1 (grainyhead-like transcription factor 1) is a regulator of desmoglein 1 (Dsg1) protein, and thus, controls differentiation of epidermis, and hair anchorage. In mice, inactivation of this gene leads to slow coat growth. It acts as a tumor suppressor in neuroblastoma, where the expression of this gene is repressed by HDAC3 (histone deacetylase) and MYCN (V-Myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70918

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomasz Wilanowski et al.
The EMBO journal, 27(6), 886-897 (2008-02-22)
In Drosophila, the grainy head (grh) gene plays a range of key developmental roles through the regulation of members of the cadherin gene family. We now report that mice lacking the grh homologue grainy head-like 1 (Grhl1) exhibit hair and
Xia Gao et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9356-9361 (2013-05-22)
Most of the airways of the human lung are lined by an epithelium made up of ciliated and secretory luminal cells and undifferentiated basal progenitor cells. The integrity of this epithelium and its ability to act as a selective barrier
Johannes Fabian et al.
Cancer research, 74(9), 2604-2616 (2014-01-15)
Neuroblastoma is an embryonic solid tumor of neural crest origin and accounts for 11% of all cancer-related deaths in children. Novel therapeutic strategies are therefore urgently required. MYCN oncogene amplification, which occurs in 20% of neuroblastomas, is a hallmark of
Saaket Varma et al.
American journal of physiology. Lung cellular and molecular physiology, 306(5), L405-L419 (2014-01-01)
Chronic injury of alveolar lung epithelium leads to epithelial disintegrity in idiopathic pulmonary fibrosis (IPF). We had reported earlier that Grhl2, a transcriptional factor, maintains alveolar epithelial cell integrity by directly regulating components of adherens and tight junctions and thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service