Skip to Content
Merck
All Photos(1)

Documents

SAB1405691

Sigma-Aldrich

Anti-CYP4B1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CYPIVB1, P-450HP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~59.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP4B1(1580)

General description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined. Two transcript variants encoding slightly different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CYP4B1 (AAH17758.1, 1 a.a. ~ 512 a.a) full-length human protein.

Sequence
MVPSFLSLSFSSLGLWASGLILVLGFLKLIHLLLRRQTLAKAMDKFPGPPTHWLFGHALEIQETGSLDKVVSWAHQFPYAHPLWFGQFIGFLNIYEPDYAKAVYSRGDPKAPDVYDFFLQWIGRGLLVLEGPKWLQHRKLLTPGFHYDVLKPYVAVFTESTRIMLDKWEEKAREGKSFDIFCDVGHMALNTLMKCTFGRGDTGLGHSRDSSYYLAVSDLTLLMQQRLVSFQYHNDFIYWLTPHGRRFLRACQVAHDHTDQVIRERKAALQDEKVRKKIQNRRHLDFLDILLGARDEDDIKLSDADLRAEVDTFMFEGHDTTTSGISWFLYCMALYPEHQHRCREEVREILGDQDFFQWDDLGKMTYLTMCIKESFRLYPPVPQVYRQLSKPVTFVDGRSLPAGSLISMHIYALHRNSAVWPDPEVFDSLRFSTENASKRHPFAFMPFSAGPRNCIGQQFAMSEMKVVTAMCLLRFEFSLDPSRLPIKMPQLVLRSKNGFHLHLKPLGPGSGK

Biochem/physiol Actions

CYP4B1 (cytochrome P450, family 4, subfamily B, polypeptide 1) plays an important role in wound healing and inflammation in the ocular surface by increasing the production of the inflammatory 12-hydroxyeicosanoids in the corneal epithelium. It is also involved in the metabolism of several xenobiotics such as 2-aminofluorene, 2-naphthylamine and benzidine.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takamitsu Sasaki et al.
Japanese journal of clinical oncology, 38(9), 634-640 (2008-08-21)
Cytochrome P450 4B1 (CYP4B1) is involved in the metabolism of several xenobiotics, such as 2-aminofluorene, 2-naphthylamine and benzidine. CYP4B1 allelic variants CYP4B1*1-*7 were recently identified. We thus hypothesized that CYP4B1 genotypes may modify bladder cancer risk. We examined the CYP4B1
Silvia Ashkar et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 20(1), 65-74 (2004-03-10)
Injury to the ocular surface provokes an inflammatory response that is mediated, at least in part, by corneal epithelial derived 12-hydroxyeicosanoids (HETEs) including 12-HETE and 12-HETrE; both metabolites exhibit potent inflammatory and angiogenic properties and are formed by a cytochrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service