Skip to Content
Merck
All Photos(2)

Documents

HPA004331

Sigma-Aldrich

Anti-CYP4B1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CYPIVB1 antibody produced in rabbit, Anti-Cytochrome P450 4B1 antibody produced in rabbit, Anti-P450-HP antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

TGSLDKVVSWAHQFPYAHPLWFGQFIGFLNIYEPDYAKAVYSRGDPKAPDVYDFFLQWIGRGLLVLEGPKWLQHRKLLTPGFHYDVLKPYVAVFTESTRIMLDKWEEKAREGKSFDIFCDVGH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP4B1(1580)

General description

CYP4B1 (cytochrome P450, family 4, subfamily B, polypeptide 1) is a member of extra-hepatic CYP450 family highly expressed in the lung. More specifically it is observed in the ciliated sub-mucosal gland ducts as well as in the surface epithelium.

Immunogen

Cytochrome P450 4B1 recombinant protein epitope signature tag (PrEST)

Application

Anti-CYP4B1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CYP4B1 (cytochrome P450, family 4, subfamily B, polypeptide 1) plays an important role in wound healing and inflammation in the ocular surface by increasing the production of the inflammatory 12-hydroxyeicosanoids in the corneal epithelium. It has high impact on the activation of environmental chemicals associated with breast cancer genesis. Various xenobiotics such as 2-aminofluorene, 2-naphthylamine and benzidine are metabolised by CYP4B1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85168

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony J Fischer et al.
American journal of respiratory cell and molecular biology, 40(2), 189-199 (2008-08-16)
Human conducting airways contain two anatomically distinct epithelial cell compartments: surface epithelium and submucosal glands (SMG). Surface epithelial cells interface directly with the environment and function in pathogen detection, fluid and electrolyte transport, and mucus elevation. SMG secrete antimicrobial molecules
M Iscan et al.
Breast cancer research and treatment, 70(1), 47-54 (2002-01-05)
Environmental chemicals are one of the risk factors in breast cancer genesis. Cytochrome P450 (CYP) enzymes play a major role in the activation of these chemicals. Using highly specific and sensitive reverse transcriptase-polymerase chain reaction (RT-PCR) analysis. the expression profile
Takamitsu Sasaki et al.
Japanese journal of clinical oncology, 38(9), 634-640 (2008-08-21)
Cytochrome P450 4B1 (CYP4B1) is involved in the metabolism of several xenobiotics, such as 2-aminofluorene, 2-naphthylamine and benzidine. CYP4B1 allelic variants CYP4B1*1-*7 were recently identified. We thus hypothesized that CYP4B1 genotypes may modify bladder cancer risk. We examined the CYP4B1
Silvia Ashkar et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 20(1), 65-74 (2004-03-10)
Injury to the ocular surface provokes an inflammatory response that is mediated, at least in part, by corneal epithelial derived 12-hydroxyeicosanoids (HETEs) including 12-HETE and 12-HETrE; both metabolites exhibit potent inflammatory and angiogenic properties and are formed by a cytochrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service