S9256
Sulfanilic acid azochromotrop
≥80% purity, powder
Synonym(s):
1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6-disulfonic acid trisodium salt, 2-(4-Sulfophenylazo)-1,8-dihydroxy-3,6-naphthalene disulfonic acid trisodium salt, 2-(4-Sulfophenylazo)chromotropic acid trisodium salt, SPADNS
About This Item
Recommended Products
Product Name
Sulfanilic acid azochromotrop, ≥80%
Assay
≥80%
form
powder
composition
Carbon: 26.0-35.0%
Nitrogen: 3.6-5.9%
technique(s)
titration: suitable
color
dark red
solubility
water: 10 mg/mL
λmax
505-510 nm
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Na+].[Na+].[Na+].Oc1cc(cc2cc(c(\N=N\c3ccc(cc3)S([O-])(=O)=O)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C16H12N2O11S3.3Na/c19-12-7-11(31(24,25)26)5-8-6-13(32(27,28)29)15(16(20)14(8)12)18-17-9-1-3-10(4-2-9)30(21,22)23;;;/h1-7,19-20H,(H,21,22,23)(H,24,25,26)(H,27,28,29);;;/q;3*+1/p-3/b18-17+;;;
InChI key
AEHGQXKBQBMQEK-GLCFPVLVSA-K
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service