Skip to Content
Merck
All Photos(4)

Documents

215309

Sigma-Aldrich

Sodium fluoride

99.99% trace metals basis

Synonym(s):

Floridine, Sodium monofluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050104
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1.4 mmHg ( 0 °C)

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

refractive index

n20/D 1.3252 (lit.)

bp

1704 °C/1 atm (lit.)

mp

993 °C (lit.)

solubility

ethanol: insoluble(lit.)

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Starting material for solid state preparation of Na3MnF6 for further high pressure structural determination studies.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlson, S. et al.
Inorganic Chemistry, 37, 1486-1486 (1998)
Naoomi Tominaga et al.
Nature communications, 6, 6716-6716 (2015-04-02)
Brain metastasis is an important cause of mortality in breast cancer patients. A key event during brain metastasis is the migration of cancer cells through blood-brain barrier (BBB). However, the molecular mechanism behind the passage through this natural barrier remains
Alexander David Ardianrama et al.
Journal of colloid and interface science, 552, 485-493 (2019-06-04)
Although triangular silver (Ag) nanoplates are intrinsically unstable, this characteristic has been taken advantage of in the development of a novel sensing platform. However, most of these applications have relied on halide ions as etchants. In the current work, we
Ryan D Espy et al.
Analytical chemistry, 86(15), 7712-7718 (2014-06-28)
Determination of eight drugs of abuse in blood has been performed using paper spray or extraction spray mass spectrometry in under 2 min with minimal sample preparation. A method has been optimized for quantification of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxy-N-methylamphetamine
E E Vincent et al.
Oncogene, 34(28), 3627-3639 (2014-09-23)
As a sensor of cellular energy status, the AMP-activated protein kinase (AMPK) is believed to act in opposition to the metabolic phenotypes favored by proliferating tumor cells. Consequently, compounds known to activate AMPK have been proposed as cancer therapeutics. However

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service