Skip to Content
Merck
All Photos(1)

Documents

S6319

Sigma-Aldrich

Sertraline hydrochloride

≥98% (HPLC)

Synonym(s):

Sertraline HCl, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NCl2 · HCl
CAS Number:
Molecular Weight:
342.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +33 to +43°, c = 2.0 in methanol

storage condition

desiccated

color

white

solubility

DMSO: >20 mg/mL

SMILES string

ClC1=C(Cl)C=CC([C@H]2C3=C(C=CC=C3)[C@@H](NC)CC2)=C1.[H]Cl

InChI

1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1

InChI key

BLFQGGGGFNSJKA-XHXSRVRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The effect of exposure of sertraline hydrochloride was studied in chronically prepared ovine fetus. It was used in glucocorticoid function assay to study the effect of antidepressants on glucocorticoid receptor function in humans.

Biochem/physiol Actions

Sertraline suppresses platelet function and exhibits antiplatelet and endothelium protective functions. It relieves depression and improves the quality of life in patients with cardiovascular disorders. It can also be prescribed to treat depression in Parkinson′s disease.
Selective serotonin reuptake inhibitor, antidepressant.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael R Domingues et al.
Toxics, 10(1) (2022-01-21)
Selective serotonin reuptake inhibitors (SSRI) are the most common antidepressant used by pregnant women; however, they have been associated with adverse pregnancy outcomes and perinatal morbidity in pregnant women and animal models. We investigated the effects of two SSRI, fluoxetine
Hannah P Fricke et al.
Frontiers in pharmacology, 14, 1244580-1244580 (2023-12-11)
Postpartum depression affects many individuals after parturition, and selective serotonin reuptake inhibitors (SSRIs) are often used as the first-line treatment; however, both SSRIs and lactation are independently associated with bone loss due to the role of serotonin in bone remodeling.
Elien Heylen et al.
British journal of cancer, 128(10), 1862-1878 (2023-03-19)
One-third of cancers activate endogenous synthesis of serine/glycine, and can become addicted to this pathway to sustain proliferation and survival. Mechanisms driving this metabolic rewiring remain largely unknown. NKX2-1 overexpressing and NKX2-1 knockdown/knockout T-cell leukaemia and lung cancer cell line
Celeste M Sheftel et al.
Physiological reports, 10(5), e15204-e15204 (2022-03-03)
Women mobilize up to 10% of their bone mass during lactation to provide milk calcium. About 8%-13% of mothers use selective serotonin reuptake inhibitors (SSRI) to treat peripartum depression, but SSRIs independently decrease bone mass. Previously, peripartal use of the
Cassidy Delaney et al.
American journal of physiology. Lung cellular and molecular physiology, 301(6), L937-L944 (2011-09-13)
Maternal use of selective serotonin (5-HT) reuptake inhibitors (SSRIs) is associated with an increased risk for persistent pulmonary hypertension of the newborn (PPHN), but little is known about 5-HT signaling in the developing lung. We hypothesize that 5-HT plays a

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service