Skip to Content
Merck
All Photos(1)

Documents

S2667

Sigma-Aldrich

Sequa-brene

gelatinous solid

Synonym(s):

Hexadimethrine bromide, Polybrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116012
NACRES:
NA.26

form

gelatinous solid

Quality Level

packaging

vial of 75 mg Sequa-brene (5 mg NaCl added)

storage temp.

2-8°C

InChI

1S/C10H24N2.C3H6Br2/c1-11(2)9-7-5-6-8-10-12(3)4;4-2-1-3-5/h5-10H2,1-4H3;1-3H2

InChI key

KZKAYEGOIJEWQB-UHFFFAOYSA-N

Application

Used as a protein/peptide carrier for protein sequencing.

Reconstitution

See data sheet for preparation.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hewan A Belete et al.
PloS one, 6(11), e27469-e27469 (2011-11-17)
Cell wounding is an important driver of the innate immune response of ventilator-injured lungs. We had previously shown that the majority of wounded alveolus resident cells repair and survive deformation induced insults. This is important insofar as wounded and repaired
Samantha J Stehbens et al.
Nature cell biology, 16(6), 561-573 (2014-05-27)
Turnover of integrin-based focal adhesions (FAs) with the extracellular matrix (ECM) is essential for coordinated cell movement. In collectively migrating human keratinocytes, FAs assemble near the leading edge, grow and mature as a result of contractile forces and disassemble underneath
[Chemiluminescence of peritoneal macrophages activated by non-natural polyelectrolytes].
L G Korkina et al.
Doklady Akademii nauk SSSR, 282(1), 206-209 (1985-05-01)
L Bonsignore et al.
Leukemia, 31(3), 614-624 (2016-08-20)
Primary effusion lymphoma (PEL) is an incurable malignancy that develops in immunodeficient patients as a consequence of latent infection of B-cells with Kaposi's sarcoma-associated herpes virus (KSHV). Malignant growth of KSHV-infected B cells requires the activity of the transcription factor
Nicolas Christoforou et al.
Scientific reports, 7, 40285-40285 (2017-01-11)
Transdifferentiation has been described as a novel method for converting human fibroblasts into induced cardiomyocyte-like cells. Such an approach can produce differentiated cells to study physiology or pathophysiology, examine drug interactions or toxicities, and engineer cardiac tissues. Here we describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service