Skip to Content
Merck
All Photos(3)

Documents

S1647

Sigma-Aldrich

(−)-Sinigrin hydrate

≥98% (HPLC), from horseradish

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16KNO9S2 · xH2O
CAS Number:
Molecular Weight:
397.46 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

horseradish

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

mp

128 (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

cation traces

K: 8.4-10.8% (anhydrous)

storage temp.

room temp

SMILES string

[K+].[H]O[H].OC[C@H]1O[C@@H](S\C(CC=C)=N\OS([O-])(=O)=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C10H17NO9S2.K.H2O/c1-2-3-6(11-20-22(16,17)18)21-10-9(15)8(14)7(13)5(4-12)19-10;;/h2,5,7-10,12-15H,1,3-4H2,(H,16,17,18);;1H2/q;+1;/p-1/b11-6+;;/t5-,7-,8+,9-,10+;;/m1../s1

InChI key

IUBVMJHASFBYGW-WBMBWNLZSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A β-D-thioglucopyranoside occurring in black mustard seeds and horseradish root. Substrate for thioglucosidase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Salehin et al.
Nature communications, 10(1), 4021-4021 (2019-09-08)
A detailed understanding of abiotic stress tolerance in plants is essential to provide food security in the face of increasingly harsh climatic conditions. Glucosinolates (GLSs) are secondary metabolites found in the Brassicaceae that protect plants from herbivory and pathogen attack.
Tahereh Aghajanzadeh et al.
Frontiers in plant science, 5, 704-704 (2015-01-08)
Brassica juncea seedlings contained a twofold higher glucosinolate content than B. rapa and these secondary sulfur compounds accounted for up to 30% of the organic sulfur fraction. The glucosinolate content was not affected by H2S and SO2 exposure, demonstrating that
Gui-xiao La et al.
Journal of Zhejiang University. Science. B, 10(6), 454-464 (2009-06-03)
The effects of CO(2) enrichment on the growth and glucosinolate (GS) concentrations in the bolting stem of Chinese kale (Brassica alboglabra L.) treated with three nitrogen (N) concentrations (5, 10, and 20 mmol/L) were investigated. Height, stem thickness, and dry
Monika Okulicz
Plant foods for human nutrition (Dordrecht, Netherlands), 65(3), 217-224 (2010-09-03)
Sinigrin (SIN) and allyl isothiocyanate (AITC) are compounds found in high concentrations in Brassica family vegetables, especially in Brussels sprouts. Recently, they have been used as a nutrition supplement for their preventive and medicinal effect on some types of cancer
Richard A Lankau
Proceedings of the National Academy of Sciences of the United States of America, 109(28), 11240-11245 (2012-06-27)
Although reciprocal evolutionary responses between interacting species are a driving force behind the diversity of life, pairwise coevolution between plant competitors has received less attention than other species interactions and has been considered relatively less important in explaining ecological patterns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service