Skip to Content
Merck
All Photos(1)

Key Documents

W203408

Sigma-Aldrich

Allyl isothiocyanate

≥95%, FCC

Synonym(s):

AITC, Oil of mustard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NCS
CAS Number:
Molecular Weight:
99.15
FEMA Number:
2034
Beilstein:
773748
EC Number:
Council of Europe no.:
2110
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.025
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.529 (lit.)

bp

150 °C (lit.)

mp

−80 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

horseradish; pungent

storage temp.

2-8°C

SMILES string

C=CCN=C=S

InChI

1S/C4H5NS/c1-2-3-5-4-6/h2H,1,3H2

InChI key

ZOJBYZNEUISWFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Allyl isothiocyanate is mainly found in Brassica species such as mustard, cabbage, and broccoli. It is formed by the degradation of the glucosinolate, sinigrin.

Biochem/physiol Actions

Naturally occurring constituent of cruciferous vegetables.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitization and desensitization to allyl isothiocyanate (mustard oil) in the nasal cavity.
Brand G & Jacquot L.
Chemical Senses, 27(7), 593-598 (2002)
Antibacterial mechanism of allyl isothiocyanate.
Lin CM, et al.
Journal of Food Protection, 63(6), 727-734 (2000)
Dong Xiao et al.
Carcinogenesis, 24(5), 891-897 (2003-05-29)
Dietary isothiocyanates (ITCs) are highly effective in affording protection against chemically induced cancers in laboratory animals. In the present study, we demonstrate that allyl isothiocyanate (AITC), a constituent of cruciferous vegetables, significantly inhibits proliferation of cultured PC-3 (androgen-independent) and LNCaP
Yuesheng Zhang
Molecular nutrition & food research, 54(1), 127-135 (2009-12-05)
Allyl isothiocyanate (AITC), which occurs in many common cruciferous vegetables, is widely and often frequently consumed by humans. Besides antimicrobial activity against a wide spectrum of pathogens, it showed anticancer activity in both cultured cancer cells and animal models, although
Meng Zhao et al.
Molecular pain, 8, 55-55 (2012-07-31)
Oxaliplatin, a platinum-based chemotherapeutic agent, causes an unusual acute peripheral neuropathy. Oxaliplatin-induced acute peripheral neuropathy appears in almost all patients rapidly after infusion, and is triggered or exacerbated by cold, while its mechanisms are poorly understood. In this study, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service