Skip to Content
Merck
All Photos(1)

Key Documents

RAB0803

Sigma-Aldrich

Mouse IgG2a ELISA Kit

Synonym(s):

IgG2a, Immunoglobulin 2a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

Mouse IgG2a enzyme-linked immunosorbent assay (ELISA) Kit is an in vitro ELISA kit for the quantitative measurement of mouse IgG2a in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Hagelkruys et al.
The Journal of experimental medicine, 218(1) (2020-09-16)
Jagunal homolog 1 (JAGN1) has been identified as a critical regulator of neutrophil biology in mutant mice and rare-disease patients carrying JAGN1 mutations. Here, we report that Jagn1 deficiency results in alterations in the endoplasmic reticulum (ER) of antibody-producing cells
Christiane S Hampe et al.
Immunity, inflammation and disease, 8(3), 299-309 (2020-05-08)
Beta-cell autoantibodies are established markers of autoimmunity, which we compared between Ghanaian adults with or without diabetes, living in rural and urban Ghana and in three European cities. In the multicenter cross-sectional Research on Obesity and Diabetes among African Migrants
Jamie Bernadette A Sy et al.
PloS one, 15(5), e0233047-e0233047 (2020-05-12)
Fruits have been widely considered as the default "health foods" because they contain numerous vitamins and minerals needed to sustain human health. Fermentation strategies have been utilized to enhance the nutritive and flavor features of healthy and readily consumable fruit
Stanislav Khil'chenko et al.
Frontiers in medicine, 7, 472-472 (2020-10-06)
Pemphigus and pemphigoid diseases are organ-specific autoimmune diseases of the skin and/or mucous membranes, which are caused by autoantibodies targeting structural proteins of the skin. In other autoimmune diseases, a high prevalence of primary antibody deficiencies was noted. Conversely, a
Haiyin Liu et al.
Journal of immunology (Baltimore, Md. : 1950), 205(5), 1207-1216 (2020-08-05)
MHC class II (MHC II) displays peptides at the cell surface, a process critical for CD4+ T cell development and priming. Ubiquitination is a mechanism that dictates surface MHC II with the attachment of a polyubiquitin chain to peptide-loaded MHC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service