Skip to Content
Merck
All Photos(1)

Key Documents

RAB0736

Sigma-Aldrich

Human SDC1 / Syndecan-1 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 14 pg/mL
standard curve range: 16.38-4000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SDC1(6382)

General description

The antibody pair provided in this kit recognize human Syndecan-1/CD138.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josephine Koch et al.
American journal of physiology. Renal physiology, 316(1), F121-F127 (2018-11-01)
Syndecan-1, a transmembrane heparan sulfate proteoglycan, associates with renal and cardiovascular functioning. We earlier reported syndecan-1 to be involved in renal tubular regeneration. We now examined plasma values of syndecan-1 in a hemodialysis cohort and its association with volume and
Lawrence N Diebel et al.
American journal of surgery, 217(6), 1076-1082 (2019-01-13)
Hyperglycemia promotes vascular inflammation; however its effect on endothelial dysfunction in sepsis is unknown. Microfluidic devices (MFD) may closely mimic the in vivo endothelial cell microenvironment. We hypothesized that stress glucose concentrations would increase sepsis related endothelial injury/activation. Human umbilical vein
Marcella C A Müller et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 30(5), 181-187 (2019-06-04)
: Coagulopathy has a high incidence in critically ill patients and is often caused by disseminated intravascular coagulation (DIC). Although the clinical picture of DIC ranges from a prothrombotic state to severe consumption coagulopathy with an increased bleeding tendency, there
D Yablecovitch et al.
Digestive diseases and sciences, 62(3), 755-760 (2016-12-28)
Syndecan-1 (SDC1) is essential for maintaining normal epithelial barrier. Shedding of SDC1 ectodomain, reflected by serum soluble syndecan-1 (SSDC1) levels, is regulated by inflammation. Increased intestinal permeability plays a central role in celiac disease (CD). The association between SSDC1 levels
Xuan Huang et al.
Frontiers in cell and developmental biology, 8, 599-599 (2020-08-08)
Mammographic density (MD) is a strong and independent factor for breast cancer (BC) risk and is increasingly associated with BC progression. We have previously shown in mice that high MD, which is characterized by the preponderance of a fibrous stroma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service