Skip to Content
Merck
All Photos(1)

Key Documents

RAB0540

Sigma-Aldrich

Human FGF19 / Fibroblast Growth Factor 19 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type urine
sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 30 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF19(9965)

General description

This ELISA antibody pair detects human Fibroblast Growth Factor 19.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Fibroblast growth factor (FGF) 19 controls bile acid, fatty acid, glucose and phosphate metabolism in target organs by activating FGF receptor 4. FGF19 regulates hepatic protein and glycogen metabolism via activation of insulin-independent endocrine pathway. In diabetic mice, FGF19 reduces the levels of serum glucose and triglycerides. Overexpression of the gene leads to the development of hepatocellular carcinoma (HCC).

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangdong Zhao et al.
Molecular carcinogenesis, 57(11), 1616-1625 (2018-08-04)
Although genetic amplification and overexpression of the fibroblast growth factor 19 (FGF19) gene are found in human breast cancer, mechanisms that contribute to such functional alterations remain elusive. We report here that high expression of FGF19 is associated with the
FGF19 as a Postprandial, Insulin-Independent Activator of Hepatic Protein and Glycogen Synthesis
Kir S, et al.
Science, 331(6024), 1621-1624 (2011)
Aze Wilson et al.
Scientific reports, 10(1), 11725-11725 (2020-07-18)
The farnesoid X receptor (FXR) is implicated in Crohn's disease (CD) pathogenesis. It is unclear how genetic variation in FXR impacts CD severity versus genetic variation in nuclear receptors such as pregnane X receptor (PXR) and the multi-drug resistance protein
Tissue-specific Expression of βKlotho and Fibroblast Growth Factor (FGF) Receptor Isoforms Determines Metabolic Activity of FGF19 and FGF21.
Kurosu H, et al.
The Journal of Biological Chemistry, 282(37), 26687-26695 (2007)
Christian Borup et al.
The American journal of gastroenterology, 115(12), 2086-2094 (2020-08-03)
The diagnosis of bile acid diarrhea is often missed because the availability of the seleno-taurohomocholic acid (SeHCAT) test is limited. We aimed to compare the biomarkers 7α-hydroxy-4-cholesten-3-one (C4) and fibroblast growth factor 19 (FGF19) with the SeHCAT test. Patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service