RAB0206
Human GH ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type plasma
sample type serum
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 2.5-600 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... GH1(2688)
General description
The Human GH ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human GH in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human GH
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aimilios Kaklamanos et al.
Scientific reports, 7(1), 2397-2397 (2017-05-27)
The human growth hormone (hGH) minigene used for transgene stabilization in mice has been recently identified to be locally expressed in the tissues where transgenes are active and associated with phenotypic alterations. Here we extend these findings by analyzing the
Yang Song et al.
Drug delivery and translational research, 8(2), 450-460 (2017-03-23)
The epidermal skin barrier plays an important role in protecting underlying structures. It allows the passage of low molecular weight lipophilic molecules, but restricts the passage of hydrophilic molecules and macromolecules. The objective of this study was to investigate the
Wei He et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(2), 792-803 (2017-11-28)
Aberrant expression of miRNA has been found in many tumor tissues to regulate the tumorigenesis by binding to the 3`- untranslated region (3`-UTR) of the target genes. The aim of this study is to investigate the role of miR-148b, miR-152/ALCAM
Takahiro Nemoto et al.
Scientific reports, 10(1), 1339-1339 (2020-01-30)
If sufficient nutrition is not obtained during pregnancy, the fetus changes its endocrine system and metabolism to protect the brain, resulting in a loss of body size. The detailed mechanisms that determine the success or failure of growth catch-up are
Sajjad Muhammad et al.
Neuromodulation : journal of the International Neuromodulation Society, 21(1), 31-37 (2017-10-25)
In our previous study, anti-inflammatory IL-10 serum levels were significantly elevated after burst spinal cord stimulation (SCS) in back pain patients and correlated with pain intensity. This current study extended cytokine analysis including metabolic-associated adipokine/cytokine serum assessment in chronic back
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service