Skip to Content
Merck
All Photos(2)

Key Documents

RAB0117

Sigma-Aldrich

Mouse KC / CXCL1 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 0.614-150 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Cxcl1(14825)

General description

The Mouse KC (keratinocyte chemoattractant; keratinocyte- derived chemokine) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse KC in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse KC

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

All three isoforms of growth-related protein (GRO) are CXC chemokines that can signal through C-X-C motif chemokine receptors 1 or 2 (CXCR1 or 2). The GRO proteins chemoattract and activate neutrophils and basophils. CXCL1 has a role in host defense against polymicrobial sepsis, upregulation of intercellular adhesion molecule 1 (ICAM-1) and migration of neutrophils. The protein also activates mitogen-activated protein kinases (MAPKs), pro-inflammatory proteins and nuclear factor-κB (NF-κB).

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth Schneider et al.
Neurology(R) neuroimmunology & neuroinflammation, 8(1) (2020-11-12)
To investigate cross-sectional associations of CSF levels of neurofilament light chain (NfL) and of the newly emerging marker chitinase 3-like protein 1 (CHI3L1) with brain and spinal cord atrophy, which are established MRI markers of disease activity in MS, to
Emily Lin et al.
Clinical & translational immunology, 8(4), e1045-e1045 (2019-04-19)
Systemic sclerosis (SSc) is an autoimmune disease characterised by fibrosis, vascular dysfunction and immune dysregulation. The pathogenesis of SSc remains poorly understood, although studies have indicated a role for the innate immune response. Here, we measured serum interleukin (IL)-1α, IL-1β
Gabriela M Escalante et al.
Vaccines, 8(2) (2020-04-10)
Primary infection with Epstein-Barr virus (EBV) is associated with acute infectious mononucleosis, whereas persistent infection is associated with chronic diseases such as autoimmune diseases and various types of cancer. Indeed, approximately 2% of all new cancer cases occurring annually worldwide
CXCL1 contributes to host defense in polymicrobial sepsis via modulating T cell and neutrophil functions.
Jin L
Journal of Immunology, 193(7), 3549-3558 (2014)
Barbara Choromańska et al.
Journal of clinical medicine, 9(4) (2020-04-05)
This is the first study to evaluate both the antioxidant barrier, glutathione metabolism, and oxidative damage to proteins and lipids in morbidly obese patients undergoing bariatric treatment. The study included 65 patients with class 3 obesity divided into two subgroups:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service