Skip to Content
Merck
All Photos(1)

Documents

ECM508

Sigma-Aldrich

QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), colorimetric

The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with colorimetric detection.

Synonym(s):

cell migration assay, colorimetric chemotaxis assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®
QCM

technique(s)

activity assay: suitable
cell based assay: suitable

detection method

colorimetric

shipped in

wet ice

Application

Research Category
Cell Structure
The CHEMICON QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. Each Chemicon Cell Migration Assay Kit contains sufficient reagents for the evaluation of 24 samples. The quantitative nature of this assay is especially useful for screening of pharmacological agents.

The CHEMICON QCM 24-well Migration Assay is intended for research use only; not for diagnostic applications.

Packaging

24 wells

Components

Sterile 24-well Cell Migration Plate Assembly: (Part No. 90333) Two 24-well plates with 12 inserts per plate (24 inserts total/kit).

Cell Stain: (Part No. 90144) One 20 mL bottle.

Extraction Buffer: (Part No. 90145) One 20 mL bottle.

Cotton Swabs: (Part No. 10202) 50 each.

Forceps: (Part No. 10203) One each.

Storage and Stability

Store kit materials at 2-8°C for up to their expiration date. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma rich in growth factors (PRGF-Endoret) stimulates proliferation and migration of primary keratocytes and conjunctival fibroblasts and inhibits and reverts style=background-c
Anitua E, Sanchez M, Merayo-Lloves J, De la Fuente M, Muruzabal F, Orive G
Investigative Ophthalmology & Visual Science null
Tieqiang Zhao et al.
American journal of physiology. Heart and circulatory physiology, 304(12), H1719-H1726 (2013-04-16)
Platelet-derived growth factor (PDGF)-D is a newly recognized member of the PDGF family with its role just now being understood. Our previous study shows that PDGF-D and its receptors (PDGFR-β) are significantly increased in the infarcted heart, where PDGFR-β is
Su Jin Hwang et al.
Scientific reports, 6, 21596-21596 (2016-02-18)
Brain metastasis is the most common type of intracranial cancer and is the main cause of cancer-associated mortality. Brain metastasis mainly originates from lung cancer. Using a previously established in vitro brain metastatic model, we found that brain metastatic PC14PE6/LvBr4
The L6 domain tetraspanin Tm4sf4 regulates endocrine pancreas differentiation and directed cell migration.
Keith R Anderson,Ruth A Singer,Dina A Balderes,Laura Hernandez-Lagunas et al.
Development null
Hon Wai Koon et al.
The American journal of pathology, 177(5), 2300-2309 (2010-10-05)
Substance P (SP) and the neurokinin-1 receptor (NK-1R) are involved in the development of colitis and mucosal healing after colonic inflammation. We studied whether SP modulates colonic fibrosis by using a chronic model of trinitrobenzenesulfonic acid (TNBS)-induced colitis in wild-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service