Skip to Content
Merck
All Photos(1)

Key Documents

R8129

Sigma-Aldrich

Renin Substrate Tetradecapeptide porcine

≥97% (HPLC), suitable for LC/MS and MALDI-TOF

Synonym(s):

Angiotensinogen 1-14, Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-leu-Val-Tyr-Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C85H123N21O20
CAS Number:
Molecular Weight:
1759.01
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Renin Substrate Tetradecapeptide porcine, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

powder

technique(s)

LC/MS: suitable
MALDI-TOF: suitable

color

white

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(N)CC(O)=O)C(C)C)C(=O)NC(Cc2cnc[nH]2)C(=O)N3CCCC3C(=O)NC(Cc4ccccc4)C(=O)NC(Cc5cnc[nH]5)C(=O)NC(CC(C)C)C(=O)NC(CC(C)C)C(=O)NC(C(C)C)C(=O)NC(Cc6ccc(O)cc6)C(=O)NC(CO)C(O)=O

InChI

1S/C85H123N21O20/c1-11-48(10)70(105-78(119)62(35-51-23-27-55(109)28-24-51)100-80(121)68(46(6)7)103-72(113)57(19-15-29-91-85(87)88)94-71(112)56(86)38-67(110)111)82(123)101-64(37-53-40-90-43-93-53)83(124)106-30-16-20-66(106)79(120)98-60(33-49-17-13-12-14-18-49)74(115)97-63(36-52-39-89-42-92-52)76(117)95-58(31-44(2)3)73(114)96-59(32-45(4)5)77(118)104-69(47(8)9)81(122)99-61(34-50-21-25-54(108)26-22-50)75(116)102-65(41-107)84(125)126/h12-14,17-18,21-28,39-40,42-48,56-66,68-70,107-109H,11,15-16,19-20,29-38,41,86H2,1-10H3,(H,89,92)(H,90,93)(H,94,112)(H,95,117)(H,96,114)(H,97,115)(H,98,120)(H,99,122)(H,100,121)(H,101,123)(H,102,116)(H,103,113)(H,104,118)(H,105,119)(H,110,111)(H,125,126)(H4,87,88,91)

InChI key

XJFQCYIFOWHHFN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wiliam A Prado et al.
Brain research, 972(1-2), 207-215 (2003-04-25)
The intracerebroventricular administration of renin substrate or angiotensin II evokes antinociception in rodents, but the brain sites where most of the renin-angiotensin system peptides act are not yet known. This study describes the antinociceptive effects of microinjecting porcine renin substrate
Endang Purwantini et al.
PloS one, 8(12), e81985-e81985 (2013-12-19)
The ability of Mycobacterium tuberculosis to manipulate and evade human immune system is in part due to its extraordinarily complex cell wall. One of the key components of this cell wall is a family of lipids called mycolic acids. Oxygenation
K A Rolls et al.
Clinical and experimental pharmacology & physiology, 21(3), 227-230 (1994-03-01)
1. Since plasma renin activity is increased in cyclosporin A (CsA)-induced hypertension in the rat, the role of the vascular renin-angiotensin system (RAS) in CsA-induced hypertension was investigated in rat mesenteric resistance vessels. 2. Female Wistar rats received CsA (10
S Kvist et al.
European journal of pharmacology, 198(1), 77-83 (1991-05-30)
The responses to angiotensinogen, angiotensinogen-(1-14) (tetradecapeptide, TDP), angiotensin I, and angiotensin II and the effect of the renin inhibitor, CH-66, the angiotensin-converting enzyme (ACE) inhibitor, perindopril, and the receptor antagonist, saralasin, were investigated in isolated femoral resistance arteries of the
Douglas C Woodhams et al.
BMC veterinary research, 8, 197-197 (2012-10-24)
While emerging diseases are affecting many populations of amphibians, some populations are resistant. Determining the relative contributions of factors influencing disease resistance is critical for effective conservation management. Innate immune defenses in amphibian skin are vital host factors against a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service