Skip to Content
Merck
All Photos(1)

Documents

R7269

Sigma-Aldrich

Rhein

≥98% (HPLC), powder, cytotoxic agent

Synonym(s):

4,5-Dihydroxyanthraquinone-2-carboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid, Cassic acid, Chrysazin 3-carboxylic acid, Monorhein, Rheic acid, Rhubarb yellow

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8O6
CAS Number:
Molecular Weight:
284.22
Beilstein:
2222155
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Rhein,

mp

≥300 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1

InChI

1S/C15H8O6/c16-9-3-1-2-7-11(9)14(19)12-8(13(7)18)4-6(15(20)21)5-10(12)17/h1-5,16-17H,(H,20,21)

InChI key

FCDLCPWAQCPTKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Rhein has been used:
  • as a potassium simplex optimization medium with amino acids (KSOMaa) to induce degeneration in embryos to study its effect on the intracellular and extracellular micro-RNA (miRNA) signature
  • as a fat mass and obesity-associated protein (FTO) inhibitor to study its role in motile ciliogenesis in an m6A-dependent manner in Xenopus embryos
  • as an FTO inhibitor to infer its antiviral action in relation with the SARS-associated coronavirus 2 (SARS-CoV-2) infection

Biochem/physiol Actions

Rhein, a cytotoxic agent, exhibits tumor suppression by activating apoptosis. It is a selective inhibitor of demethylase fat mass and obesity-associated protein (FTO). Rhein is known to participate in various biological processes including obesity and adipogenesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yaron et al.
Osteoarthritis and cartilage, 7(3), 272-280 (1999-05-18)
The etiology of osteoarthritis (OA) is still a matter of debate. Several factors are known to be involved in the destruction of the articular cartilage. Interleukin-1 (IL-1) plays an important role in the pathogenesis of osteoarthritis (OA) either directly or
J P Pelletier et al.
Clinical and experimental rheumatology, 21(2), 171-177 (2003-05-16)
The primary objective of this study was to evaluate the ex vivo therapeutic efficacy of diacerein and its active metabolite, rhein, on osteoarthritic (OA) cartilage chondrocyte DNA fragmentation and death in the experimental canine model of OA. The study also
Hyunjoon Kim et al.
Developmental cell, 56(8), 1118-1130 (2021-03-25)
Adenosine N6-methylation (m6A) is one of the most pervasive mRNA modifications, and yet the physiological significance of m6A removal (demethylation) remains elusive. Here, we report that the m6A demethylase FTO functions as a conserved regulator of motile ciliogenesis. Mechanistically, FTO
Carla Zannella et al.
Pharmaceuticals (Basel, Switzerland), 14(11) (2021-11-28)
The rapid spread of SARS-CoV-2 and the resulting pandemic has led to a spasmodic search for approaches able to limit the diffusion of the disease. The epigenetic machinery has aroused considerable interest in the last decades, and much evidence has
David Connor Hawke et al.
Frontiers in genetics, 12, 655882-655882 (2021-06-15)
Extracellular microRNA (miRNA) sequences derived from the pre-implantation embryo have attracted interest for their possible contributions to the ongoing embryonic-uterine milieu, as well as their potential for use as accessible biomarkers indicative of embryonic health. Spent culture media microdroplets used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service