Skip to Content
Merck

1377803

USP

Meclofenamate Sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Meclofenamic acid sodium salt, 2-[(2,6-Dichloro-3-methylphenyl)amino]benzoic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H10Cl2NO2Na
CAS Number:
Molecular Weight:
318.13
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

meclofenamate

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

SMILES string

[Na].Cc1ccc(Cl)c(Nc2ccccc2C(O)=O)c1Cl

InChI

1S/C14H11Cl2NO2.Na.H/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19;;/h2-7,17H,1H3,(H,18,19);;

InChI key

CHGIZYUDGOIAFY-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Cyclooxygenase (IC50 = 0.6 μM) and 5-lipoxygenase (IC50 = 46.3 μM) inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.


Still not finding the right product?

Explore all of our products under Meclofenamate Sodium


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library