Skip to Content
Merck
All Photos(1)

Key Documents

R5155

Sigma-Aldrich

Rocuronium bromide

≥97% (perchloric acid titration)

Synonym(s):

(+)-Rocuronium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H53BrN2O4
CAS Number:
Molecular Weight:
609.68
MDL number:
UNSPSC Code:
51152000
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (perchloric acid titration)

form

powder

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

[Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N5CCOCC5)[N+]6(CCCC6)CC=C

InChI

1S/C32H53N2O4.BrH/c1-5-14-34(15-6-7-16-34)28-20-26-24-9-8-23-19-29(36)27(33-12-17-37-18-13-33)21-32(23,4)25(24)10-11-31(26,3)30(28)38-22(2)35;/h5,23-30,36H,1,6-21H2,2-4H3;1H/q+1;/p-1/t23-,24+,25-,26-,27-,28-,29-,30-,31-,32-;/m0./s1

InChI key

OYTJKRAYGYRUJK-FMCCZJBLSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Rocuronium Bromide is a fumarate, non-depolarizing, neuromuscular blocking agent. It is rapid-acting, relaxes the skeletal muscle and decreases oxygen consumption during surgery. It competes with acetylcholine and bind to cholinergic receptors at neuromuscular junctions.
Aminosteroid; neuromuscular blocker for anaesthesia

Preparation Note

Rocuronium bromide yields clear, light yellow solution in water at 100 mg/ml.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mats Eriksson et al.
Clinical laboratory, 61(7), 825-829 (2015-08-25)
Determination of troponin I may be important in the management of the critically ill patient. In medical emergencies, especially when vascular access is difficult to achieve, the use of intraosseous (10) needles is recommended. We aimed to perform a descriptive
Yuji Kadoi et al.
The journal of ECT, 29(1), 33-36 (2012-10-12)
This study was conducted to (1) compare the recovery times from rocuronium-induced muscle relaxation after reversal with sugammadex between young and elderly patients undergoing electroconvulsive therapy (ECT), and (2) to examine the existence of a correlation between cardiac index and
Adrienn Pongrácz et al.
Anesthesiology, 119(1), 36-42 (2013-05-15)
Doses of sugammadex required to reverse deep, moderate, and shallow rocuronium-induced neuromuscular blockade have been established. However, no adequate doses for the reversal of reappearance of four twitches of train-of-four (TOF) stimulation (threshold TOF-count-four) have been established. This single-center, randomized
T Wang et al.
Anaesthesia, 69(8), 854-859 (2014-05-14)
We investigated whether laparoscopic vs open surgical approaches affected the duration of neuromuscular blockade following a single bolus dose of rocuronium. Fifty-three female patients underwent either laparoscopic or open gynaecological surgery. Rocuronium 0.6 mg.kg(-1) was administered to achieve neuromuscular blockade
Shana L Ballow et al.
The journal of trauma and acute care surgery, 73(6), 1401-1405 (2012-11-29)
In the emergency department (ED) of a teaching hospital, rapid sequence intubation (RSI) is performed by physicians with a wide range of experience. A variety of medications have been used for RSI, with potential for inadequate or excessive dosing as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service