Skip to Content
Merck
All Photos(1)

Key Documents

PZ0039

Sigma-Aldrich

Lorlatinib

≥98% (HPLC)

Synonym(s):

(10R)-7-Amino-12-fluoro-2,10,16-trimethyl-15-oxo-10,15,16,17-tetrahydro-2H.8,4-(metheno)pyrazolo[4,3.h][2,5,11]-benzoxadiazacyclotetradecine-3-carbonitrile, PF-06463922, PF-6463922

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19FN6O2
CAS Number:
Molecular Weight:
406.41
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -95 to -115°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

NC1=C(O[C@@H](C2=C3C=CC(F)=C2)C)C=C(C4=C(C#N)N(C)N=C4CN(C)C3=O)C=N1

InChI key

IIXWYSCJSQVBQM-LLVKDONJSA-N

Biochem/physiol Actions

Lorlatinib (PF-06463922) is a potent, selective brain-penetrable inhibitor of both anaplastic lymphoma kinase (ALK) and c-ros Oncogene 1 (ROS1) with strong activity against most known ALK and ROS1 mutants identified in patients with crizotinib-resistant disease. It is in clinical trials for the treatment of non–small cell lung cancer (NSCLC).
Potent, selective inhibitor of both ALK and ROS1

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Dong et al.
BMC pulmonary medicine, 18(1), 13-13 (2018-01-25)
Crizotinib is recommended as first-line therapy in ROS1-driven lung adenocarcinoma. However, the optimal first-line therapy for this subgroup of lung cancer is controversial according to the available clinical data. Here, we describe a 57-year-old man who was diagnosed with stage
Ted W Johnson et al.
Journal of medicinal chemistry, 57(11), 4720-4744 (2014-05-14)
Although crizotinib demonstrates robust efficacy in anaplastic lymphoma kinase (ALK)-positive non-small-cell lung carcinoma patients, progression during treatment eventually develops. Resistant patient samples revealed a variety of point mutations in the kinase domain of ALK, including the L1196M gatekeeper mutation. In
Alice T Shaw et al.
The Lancet. Oncology, 18(12), 1590-1599 (2017-10-28)
Most patients with anaplastic lymphoma kinase (ALK)-rearranged or ROS proto-oncogene 1 (ROS1)-rearranged non-small-cell lung cancer (NSCLC) are sensitive to tyrosine kinase inhibitor (TKI) therapy, but resistance invariably develops, commonly within the CNS. This study aimed to analyse the safety, efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service