Skip to Content
Merck
All Photos(2)

Key Documents

MITOISO2

Sigma-Aldrich

Mitochondria Isolation Kit

sufficient for 50 applications (2-5 x 107 cells), isolation of enriched mitochondrial fraction from cells

Synonym(s):

Mitochondrial Extraction Kit, Mitochondrial Purification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

usage

sufficient for 50 applications (2-5 x 107 cells)

technique(s)

ELISA: suitable
western blot: suitable

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Mitochondria, a double membrane structure, is the site of most energy production in eukaryotic cells.[1] The mitochondria isolation kit contains two buffers: one for extracting proteins for functional studies and another for profiling in proteome studies. Additionally, the kit includes a storage buffer for preserving intact mitochondria.

Application

Mitochondria Isolation Kit has been used in the isolation of mitochondria from:
  • Ewing′s Sarcoma Family of Tumor (ESFT) cells[2]
  • blood samples of tuberculosis patients[3]
  • human neuroblastoma cells and glioma cells[4]
  • myotubes[5]
  • HepG2 cells[6]

Features and Benefits

  • The kit allows for quick and simple extraction of a concentrated portion of mitochondria from cells.
  • The isolated mitochondria will mostly have intact inner and outer membranes.
  • Can be used to extract mitochondrial proteins for proteome analysis

Other Notes

  • This product is for R&D use only, not for drug, household, or other uses.
  • Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
  • Use ultrapure water for the preparation of reagents.
  • Please refer to the product information sheet to learn more about kit components, reagents required, storage and stability, and isolation protocols.

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2978CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization. 10 mLSDS

  • P8340Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution 2.5 mLSDS

  • C0356Protein Extraction Reagent Type 4 1 bottleSDS

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youguang Gao et al.
Journal of translational medicine, 18(1), 114-114 (2020-03-07)
We have reported that polydatin (PD) alleviates mitochondrial dysfunction in rat models of sepsis-induced acute kidney injury (SI-AKI), but the mechanism is not well understood. Here, we investigated the role of Parkin-mediated mitophagy in the protective effects of PD in
Vladislav V Glinsky et al.
Neoplasia (New York, N.Y.), 11(9), 901-909 (2009-09-03)
At present, there is no efficient curative therapy for cancer patients with advanced metastatic disease. Targeting of antiapoptotic molecules acting on the mitochondrial apoptosis pathway could potentially augment antimetastatic effect of cytotoxic drugs. Similarly to Bcl-2 family members, beta-galactoside-binding lectin
Mitochondria
Cooper G M
Journal of biological and chemical chronicles (2000)
Theodora Panagaki et al.
Biomolecules, 10(4) (2020-04-29)
Down syndrome (trisomy of human chromosome 21) is a common genetic disorder. Overproduction of the gaseous mediator hydrogen sulfide (H2S) has been implicated in the pathogenesis of neurological and metabolic deficits associated with Down syndrome. Several lines of data indicate
Imran Khan et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 789-799 (2016-10-11)
Although quercetin is very well known for its anticancer activity, however it shows some drawbacks. Herein, we have evaluated the apoptotic effect TEF (5, 3'-dihydroxy-3, 7, 4'-triethoxyflavone), a newly synthesized quercetin derivative on HCT-116 colon cancer cells. After 24h of

Articles

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service