Skip to Content
Merck
All Photos(2)

Key Documents

PHG0007

SAFC

MOPS

Pharma Manufacturing

Synonym(s):

3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO4S
CAS Number:
Molecular Weight:
209.26
Beilstein:
1106776
EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥99.5%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

pH

2.5-4 (1 M in H2O)

useful pH range

6.5-7.9

pKa (25 °C)

7.2

λ

1 M in H2O

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

OS(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

MOPS is a biological buffer also referred to as a “Propane Sultone” buffer. The pKa of MOPS is 7.01 which make it an ideal candidate for medias and protein based buffer formulations to maintain a stable environment in solution. MOPS is considered to be non-toxic to culture cell lines and provides high-solution clarity.

MOPS is used in cell culture media, biopharmaceutical buffer formulations (both upstream and downstream). MOPS based buffers are used in purification bioprocesses of antibodies, peptides, proteins and blood components.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junji Suzuki et al.
Nature communications, 5, 4153-4153 (2014-06-14)
The endoplasmic reticulum (ER) and mitochondria accumulate Ca(2+) within their lumens to regulate numerous cell functions. However, determining the dynamics of intraorganellar Ca(2+) has proven to be difficult. Here we describe a family of genetically encoded Ca(2+) indicators, named calcium-measuring
Y S L Lee et al.
Human reproduction (Oxford, England), 30(3), 543-552 (2015-01-09)
What is the relationship between cleavage stage embryo kinetics, blastocyst metabolism and subsequent embryo viability? Embryos cleaving faster at the first cleavage division resulted in blastocysts with a larger inner cell mass (ICM), higher glucose consumption, lower glycolytic rate, higher
Sanna Byström et al.
Journal of proteome research, 13(11), 4607-4619 (2014-09-19)
The brain is a vital organ and because it is well shielded from the outside environment, possibilities for noninvasive analysis are often limited. Instead, fluids taken from the spinal cord or circulatory system are preferred sources for the discovery of
Alexandre Albanese et al.
ACS nano, 8(6), 5515-5526 (2014-05-07)
A nanoparticle's physical and chemical properties at the time of cell contact will determine the ensuing cellular response. Aggregation and the formation of a protein corona in the extracellular environment will alter nanoparticle size, shape, and surface properties, giving it
Sewa Rijal et al.
Blood, 125(18), 2815-2824 (2015-03-05)
Phosphoinositide signaling regulates diverse cellular functions. Phosphoinositide-3 kinase (PI3K) generates PtdIns(3,4,5)P3 and PtdIns(3,4)P2, leading to the activation of proliferative and anti-apoptotic signaling pathways. Termination of phosphoinositide signaling requires hydrolysis of inositol ring phosphate groups through the actions of PtdIns(3,4,5)P3 3-phosphatase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service