Merck
  • Affinity proteomic profiling of plasma, cerebrospinal fluid, and brain tissue within multiple sclerosis.
Affinity proteomic profiling of plasma, cerebrospinal fluid, and brain tissue within multiple sclerosis.
Journal of proteome research (2014-09-19)
Sanna Byström, Burcu Ayoglu, Anna Häggmark, Nicholas Mitsios, Mun-Gwan Hong, Kimi Drobin, Björn Forsström, Claudia Fredolini, Mohsen Khademi, Sandra Amor, Mathias Uhlén, Tomas Olsson, Jan Mulder, Peter Nilsson, Jochen M Schwenk
ABSTRACT
The brain is a vital organ and because it is well shielded from the outside environment, possibilities for noninvasive analysis are often limited. Instead, fluids taken from the spinal cord or circulatory system are preferred sources for the discovery of candidate markers within neurological diseases. In the context of multiple sclerosis (MS), we applied an affinity proteomic strategy and screened 22 plasma samples with 4595 antibodies (3450 genes) on bead arrays, then defined 375 antibodies (334 genes) for targeted analysis in a set of 172 samples and finally used 101 antibodies (43 genes) on 443 plasma as well as 573 cerebrospinal spinal fluid (CSF) samples. This revealed alteration of protein profiles in relation to MS subtypes for IRF8, IL7, METTL14, SLC30A7, and GAP43. Respective antibodies were subsequently used for immunofluorescence on human post-mortem brain tissue with MS pathology for expression and association analysis. There, antibodies for IRF8, IL7, and METTL14 stained neurons in proximity of lesions, which highlighted these candidate protein targets for further studies within MS and brain tissue. The affinity proteomic translation of profiles discovered by profiling human body fluids and tissue provides a powerful strategy to suggest additional candidates to studies of neurological disorders.
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%