Skip to Content
Merck
All Photos(1)

Key Documents

P9544

Sigma-Aldrich

Phosphoglucose Isomerase from rabbit muscle

Type XI, lyophilized powder, ≥200 units/mg protein

Synonym(s):

D-Glucose-6-phosphate ketol-isomerase, PGI, Phosphosaccharomutase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

rabbit muscle

type

Type XI

form

lyophilized powder

specific activity

≥200 units/mg protein

composition

protein, 70-100% biuret

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Phosphoglucose Isomerase (PGI) is an enzyme crucial for the interconversion of D-glucose 6-phosphate and D-fructose 6-phosphate. PGI is responsible for the second step of glycolysis and is involved in glucogenesis[1]. It is highly conserved in bacteria and eukaryotes. It is used in sugar assays to convert fructose to glucose [2]. Product 9544 is type XI and is from rabbit muscle. It is useful in enzyme systems requiring low sulfate.

Biochem/physiol Actions

Phosphoglucose Isomerase fuctions as an isomerase, neuroleukin, autocrine motility factor, and a differentiation and maturation mediator [1]. It is responsible for the photosynthetic reduction of carbon.

Unit Definition

One unit will convert 1.0 μmole of D-fructose 6-phosphate to D-glucose 6-phosphate per min at pH 7.4 at 25 °C.

Physical form

Essentially sulfate-free powder containing citrate buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B K Campbell et al.
Reproduction (Cambridge, England), 140(5), 721-732 (2010-08-19)
Glucose is a critical metabolic fuel in most mammals although many foodstuffs also contain high levels of the monosaccharides, galactose and fructose. The aims of this work were to determine the insulin response to challenges of these sugars (experiment 1)
Yunfeng Peng et al.
Physiologia plantarum, 148(4), 470-480 (2012-10-16)
Nitrogen (N) limitation reduces leaf growth and photosynthetic rates of maize (Zea mays), and constrains photosynthate translocation to developing ears. Additionally, the period from about 1 week before to 2 weeks after silking is critical for establishing the reproductive sink
C J Jeffery et al.
Biochemistry, 39(5), 955-964 (2000-02-02)
The multifunctional protein phosphoglucose isomerase, also known as neuroleukin, autocrine motility factor, and differentiation and maturation mediator, has different roles inside and outside the cell. In the cytoplasm, it catalyzes the second step in glycolysis. Outside the cell, it serves
Sean D Schoville et al.
PloS one, 7(6), e40035-e40035 (2012-07-07)
The marine copepod Tigriopus californicus lives in intertidal rock pools along the Pacific coast, where it exhibits strong, temporally stable population genetic structure. Previous allozyme surveys have found high frequency private alleles among neighboring subpopulations, indicating that there is limited
Angela Pizzolla et al.
The American journal of pathology, 183(4), 1144-1155 (2013-08-06)
Immunization with human glucose-6-phosphate isomerase (hG6PI) protein or with several of its peptides induces arthritis in DBA/1 mice. We investigated G6PI peptide-induced arthritis in C57BL/10 mice and the effect of oxidative burst on disease. To study the arthritogenicity of G6PI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service