Skip to Content
Merck
All Photos(1)

Key Documents

11426362001

Roche

Hexokinase (HK)

suspension, suitable for detection

Synonym(s):

HK, Hexokinase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

form

suspension

Quality Level

specific activity

~450 units/mg protein (At 25 °C and pH 7.6 with D-glucose and ATP as the substrates.)

packaging

pkg of 1 mL (1,500 U)

manufacturer/tradename

Roche

pH range

6.0-8.0

application(s)

detection

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

ATP:D-hexose 6-phosphotransferase
Hexokinase consists of 486 amino acids and has a molecular weight of 57,000 D (SDS-PAGE) in citrate/phosphate buffer. It may form dimers under other buffer conditions.
Hexokinase exists in three isozymes, namely type I, type II and type III that differ in subcellular localization, catalytic and regulatory properties. The type I isozyme serves a catabolic function facilitating the entry of glucose into glycolytic metabolism for energy production. The type II and type III enzymes perform anabolic functions, such as generating G6P for glycogen synthesis or metabolism through the phosphogluconate pathway for lipid synthesis. Type I and type II isozymes have been found to be bound to mitochondria. The type III isozyme has been found to have perinuclear localization in several cell types.[1]

Application

Hexokinase (HK) has been used in enzyme assays[2][3] and chemiluminescence assay.[4] It has also been used to determine ATP concentration.[5]
Use Hexokinase for the determination of D-glucose[6][2], D-fructose, and D-sorbitol in food or biological research samples. Hexokinase may also be used for the assay of other saccharides, which are convertible to glucose or fructose, and is, therefore, useful in the assay of many glycosides.
Hexokinase has been used in the enzymatic detection of nitric oxide[4] and in the measurement of ATP by luminescence method.[7]

Biochem/physiol Actions

Hexokinase catalyzes the conversion of hexoses to hexose-6-phosphate in an ATP-dependent manner. It is also called as the glycolytic enzyme as it catalyzes the first step in glucose metabolism with the formation of glucose-6-phosphate (G6P). In eukaryotes, it functions as a glucose sensor.[8]

Quality

Contaminants: <0.002% PGI, <0.0005% G6P-DH and GR, each, <0.001% CK, MK, 6-PGDH, and ADH, each, <0.05% GIDH and invertase (β-fructosidase), each, <30 μg/ml glucose (enzymatically)

Unit Definition

Unit Assay: For measuring hexokinase activity 0.22 M glucose in 0.1 M triethanolamine buffer, pH 7.6, 6.5 mM MgCI2, 2.7 mM ATP, 0.83 mM NADP is incubated in the presence of 1.7 U glucose-6-phosphate dehydrogenase with an appropriate amount of hexokinase (10–20 mU) at +25 °C (total volume: 3.0 ml). The enzyme activity is calculated from the increase of absorbance at 340 nm ( = 6.3 [I × mmol/l-1 × cm-1]) or 365 nm ( = 3.5 [I × mmol/l-1 × cm-1]).

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6.5

Preparation Note

Activator: Hexokinase requires Mg2+ ions (Km = 2.6 mM) for activity. Catecholamines too increase activity.
Stabilizers: Thiols

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogen signaling prevents diet-induced hepatic insulin resistance in male mice with obesity
Zhu L, et al.
American Journal of Physiology. Endocrinology and Metabolism, 306(10) (2014)
Stefan Hof et al.
Frontiers in medicine, 9, 867298-867298 (2022-05-17)
Acute hemorrhage results in perfusion deficit and regional hypoxia. Since failure of intestinal integrity seem to be the linking element between hemorrhage, delayed multi organ failure, and mortality, it is crucial to maintain intestinal microcirculation in acute hemorrhage. During critical
Is there a pace-of-life syndrome linking boldness and metabolic capacity for locomotion in bluegill sunfish?
Binder TR, et al.
Animal behaviour, 121, 175-183 (2016)
Ariane D Minet et al.
Biogerontology, 13(3), 277-285 (2012-02-10)
The free radical theory of aging says that increased oxidative stress and mitochondrial dysfunction are associated with old age. In the present study we have investigated the effects of cellular senescence on muscle energetic by comparing mitochondrial content and function
Sharon Kaisari et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(9) (2022-02-27)
The mitotic (or spindle assembly) checkpoint system ensures accurate chromosome segregation in mitosis by preventing the onset of anaphase until correct bipolar attachment of sister chromosomes to the mitotic spindle is attained. It acts by promoting the assembly of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service