Skip to Content
Merck
All Photos(1)

Documents

P9537

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)−R-Phycoerythrin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)−R-PE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

phycoerythrin (R-PE) conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect immunofluorescence: 1:20

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Binds all rabbit Igs.

Application

Anti-Rabbit IgG (whole molecule)−R-Phycoerythrin antibody produced in goat has been used in flow cytometry.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. Conjugation of R-Phycoerythrin to the antibody enables the use in immunofluorescent techniques.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.1 mM EDTA, 1 mM iodoacetamide, 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of CC-chemokine receptor 7 expression on murine T cells in lymphoid tissues
Bjorkdahl O, et al.
Immunology, 110(2), 170-179 (2003)
Lothar Luther Reich et al.
Journal of molecular biology, 427(11), 2135-2150 (2014-10-15)
Uncovering the relationships between peptide and protein sequences and binding properties is critical for successfully predicting, re-designing and inhibiting protein-protein interactions. Systematically collected data that link protein sequence to binding are valuable for elucidating determinants of protein interaction but are
Transcriptional profile of in vitro expanded human epidermal progenitor cells for the treatment of non-healing wounds
Langa P, et al.
Journal of Dermatological Science, 89(3), 272-281 (2018)
Nikolaos Memos et al.
Journal of neuroscience research, 89(9), 1409-1418 (2011-05-28)
The 14-3-3 family members play a crucial role in the determination of cell fate, exerting their antiapoptotic activity through directly interfering with the critical function of the mitochondrial core proapoptotic machinery. Dimerization of 14-3-3 is vital for the interaction with
Nadine Honke et al.
Biomedicines, 10(8) (2022-08-27)
The sympathetic nervous system (SNS) has either a pro-inflammatory or anti-inflammatory effect, depending on the stage of arthritis. In the past, treatment of arthritic B cells with a β2-adrenergic receptor (β2-ADR) agonist has been shown to attenuate arthritis. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service