
Anti-Rabbit IgG (whole molecule)−R-Phycoerythrin antibody produced in goat

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing