Skip to Content
Merck
All Photos(4)

Documents

P9528

Sigma-Aldrich

Polyuridylic acid potassium salt

Synonym(s):

Poly(U) potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

Assay

≥98% (TLC)

Quality Level

form

lyophilized powder

solubility

water: 19.60-20.40 mg/mL, clear to slightly hazy, colorless

storage temp.

−20°C

InChI

1S/C9H13N2O9P/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H,10,12,15)(H2,16,17,18)/t4-,6-,7-,8-/m1/s1

InChI key

DJJCXFVJDGTHFX-XVFCMESISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyuridylic acid (Poly(U)), a homopolymer of uridine, is used as a single-stranded RNA model compound often in comparison with other ssRNA model oligonucleotides such as Poly(I) and Poly(C) to study processes that are dependent upon ssRNA regulation such as Toll-like receptor regulation. Poly(U) is used as an effective agonist for TLR7.

Preparation Note

Prepared from UDP using polynucleotide phosphorylase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soren Nielsen et al.
Science (New York, N.Y.), 340(6140), 1577-1580 (2013-07-03)
Gene expression in organisms involves many factors and is tightly controlled. Although much is known about the initial phase of transcription by RNA polymerase III (Pol III), the enzyme that synthesizes the majority of RNA molecules in eukaryotic cells, termination
Liam M Longo et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(27), 15731-15739 (2020-06-21)
De novo emergence demands a transition from disordered polypeptides into structured proteins with well-defined functions. However, can polypeptides confer functions of evolutionary relevance, and how might such polypeptides evolve into modern proteins? The earliest proteins present an even greater challenge
Tiago Antonio de Souza et al.
PloS one, 7(2), e32305-e32305 (2012-03-03)
Plant pathogenic bacteria utilize an array of effector proteins to cause disease. Among them, transcriptional activator-like (TAL) effectors are unusual in the sense that they modulate transcription in the host. Although target genes and DNA specificity of TAL effectors have
Yann Fichou et al.
Frontiers in neuroscience, 13, 1339-1339 (2020-01-11)
The aggregation of the human tau protein into neurofibrillary tangles is directly diagnostic of many neurodegenerative conditions termed tauopathies. The species, factors and events that are responsible for the initiation and propagation of tau aggregation are not clearly established, even
Debipreeta Bhowmik et al.
PloS one, 7(5), e37939-e37939 (2012-06-06)
Binding of two 9-O-(ω-amino) alkyl ether berberine analogs BC1 and BC2 to the RNA triplex poly(U)(•)poly(A)(*)poly(U) was studied by various biophysical techniques. Berberine analogs bind to the RNA triplex non-cooperatively. The affinity of binding was remarkably high by about 5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service