Skip to Content
Merck
All Photos(2)

Key Documents

P8412

Sigma-Aldrich

o-Phenylenediamine dihydrochloride

chromogenic, tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3912045
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet

form

tablet

mp

258 °C (dec.) (lit.)

solubility

water: 1 tablet/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Phenylenediamine dihydrochloride is a binucleophile and its condensation with α,β-unsaturated carbonyl compounds and aldehydes yields benzimidazoles.
o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

o-Phenylenediamine dihydrochloride has been used as a substrate for horse radish peroxidase in detecting:
  • human immunoglobulin
  • CTLA4Ig in serum samples
  • albumin in culture medium

Reconstitution

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human amniotic epithelial cells are promising transgene carriers for allogeneic cell transplantation into liver
Sakuragawa N, et al.
Journal of Human Genetics, 45(3), 171-171 (2000)
Kazuo Ohnishi et al.
Japanese journal of infectious diseases, 65(1), 19-27 (2012-01-26)
The H5N1 subtype of the highly pathogenic (HP) avian influenza virus has been recognized for its ability to cause serious pandemics among humans. In the present study, new monoclonal antibodies (mAbs) against viral proteins were established for the
Acetic acid-promoted condensation of o-phenylenediamine with aldehydes into 2-aryl-1-(arylmethyl)-1H-benzimidazoles under microwave irradiation
Azarifar D, et al.
J. Serb. Chem. Soc., 75(9), 1181-1189 (2010)
Asier Sáez-Cirión et al.
Nature protocols, 5(6), 1033-1041 (2010-06-12)
To advance T cell-based HIV vaccine development, it is necessary to evaluate the immune correlates of a protective CD8(+) T-cell response. We have developed an assay that assesses the capacity ex vivo of HIV-specific CD8(+) T cells to suppress HIV-1
In vitro and in vivo properties differ among liquid intravenous immunoglobulin preparations
Dhainaut F, et al.
Vox Sanguinis, 104(2), 115-126 (2013)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service