Skip to Content
Merck
All Photos(5)

Documents

P23938

Sigma-Aldrich

o-Phenylenediamine

flaked, 99.5%

Synonym(s):

1,2-Diaminobenzene, 1,2-Phenylenediamine, OPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
606074
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 25 °C)

Assay

99.5%

form

powder

bp

256-258 °C

mp

100-102 °C

SMILES string

Nc1ccccc1N

InChI

1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2

InChI key

GEYOCULIXLDCMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

o-Phenylenediamine can be used:
  • To prepare quinoxalines, benzimidazoles, and 1H-1,5-benzodiazepines by condensing with aryl aldehydes or ketones.
  • In the H2O mediated one-pot preparation of 1,2-disubstituted benzimidazoles by reacting with aldehydes in the presence of trimethylsilyl chloride.
  • In the copper-catalyzed synthesis of alkyne substituted quinoxalines by reacting with terminal alkynes.
  • In the preparation of 1,5-benzodiazepine derivatives by reacting with different ketones under neat reaction conditions via ytterbium trichloride catalyzed condensation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of Benzimidazoles, Quinoxalines and 3, 3-Dihydro-1H-1, 5-benzodiazepines Starting from o-Phenylenediamine
Yong Cui, et al.
Chin. J. Chem., 23(5), 589-595 (2005)
Water mediated chemoselective synthesis of 1, 2-disubstituted benzimidazoles using o-phenylenediamine and the extended synthesis of quinoxalines
Wan J, et al.
Green Chemistry, 11(10), 1633-1637 (2009)
Efficient Synthesis of 1, 5-Benzodiazepine Derivatives by Ytterbium Trichloride-Catalyzed Condensation of o-Phenylenediamine and Ketones
Wu J, et al.
Synthetic Communications, 36(4), 457-464 (2006)
Hai-Long Wang et al.
Acta tropica, 137, 58-66 (2014-05-13)
Nasal vaccination is an effective therapeutic regimen for preventing certain infectious diseases. The mucosal immune response is important for resistance to Toxoplasma gondii infection. In this study, we evaluated the immune responses elicited in BALB/c mice by nasal immunisation with
Galber Rodrigues Araujo et al.
Arthritis research & therapy, 17, 168-168 (2015-06-24)
Rheumatoid arthritis (RA) is a chronic inflammatory autoimmune disease that affects around 1% of the human population worldwide. RA diagnosis can be difficult as there is no definitive test for its detection. Therefore, the aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service