Skip to Content
Merck
All Photos(1)

Key Documents

P5520

Sigma-Aldrich

Paraldehyde

≥97%

Synonym(s):

2,4,6-Trimethyl-1,3,5-trioxacyclohexane, s-Trimethyltrioxymethylene, Acetaldehyde trimer, NSC 9799, Paracetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Beilstein:
80142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

drug control

USDEA Schedule IV

density

0.994 g/mL at 20 °C (lit.)

SMILES string

CC1OC(C)OC(C)O1

InChI

1S/C6H12O3/c1-4-7-5(2)9-6(3)8-4/h4-6H,1-3H3

InChI key

SQYNKIJPMDEDEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Modified Sanford reaction
  • Synthesis of N-arylbenzoquinaldinium derivatives with antimicrobial activity
  • Hydroformylation of alkenylamines
  • Studies of molecules with inhibitory activity against HIV-1 integrase
  • Irregular Wittig reactions for the formation of α-hydroxyketones

Biochem/physiol Actions

Hypnotic with rapid onset; anticonvulsant at high doses; used to treat delirium tremors

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Ikumi et al.
Epilepsy research, 82(2-3), 215-218 (2008-09-23)
Malaria infection reduces the binding capacity of benzodiazepine receptors in mice. We studied the efficacy of diazepam terminating seizures in children with falciparum malaria. Diazepam stopped seizures in fewer patients with malaria parasitaemia (chi(2)=3.93, P=0.047) and those with clinical diagnosis
Sethu Gunasekaran et al.
Journal of molecular modeling, 14(5), 375-383 (2008-02-27)
The FTIR and Laser-Raman spectra of paraldehyde have been recorded in the regions 4000-400 cm(-1) and 3500-250 cm(-1) respectively. Molecular electronic energy, geometrical structure, harmonic vibrational spectra, infrared intensities and Raman scattering activities have been computed at the HF/6-31G(d,p) and
O E Antia-Obong et al.
Journal of tropical pediatrics, 38(4), 173-175 (1992-08-01)
In a study of 149 cases of neonatal tetanus (NNT) admitted into the University of Calabar Teaching Hospital, 49 (33 per cent) were also found to have septicaemia. The dominant organisms were coliforms and Staphylococcus aureus. A comparison of the
Use of paraldehyde in neonates - an experience.
D Mishra et al.
Indian pediatrics, 38(2), 209-210 (2001-02-27)
The role of paraldehyde and lidocaine in the management of status epilepticus.
M Thulasimani et al.
Anaesthesia, 57(1), 99-100 (2002-02-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service