Skip to Content
Merck
All Photos(1)

Key Documents

76260

Sigma-Aldrich

Paraldehyde

≥97.0% (GC)

Synonym(s):

2,4,6-Trimethyl-1,3,5-trioxacyclohexane, s-Trimethyltrioxymethylene, Acetaldehyde trimer, NSC 9799, Paracetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Beilstein:
80142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

drug control

USDEA Schedule IV

refractive index

n20/D 1.405

bp

123-124 °C

density

0.994 g/mL at 20 °C (lit.)

functional group

ether

SMILES string

CC1OC(C)OC(C)O1

InChI

1S/C6H12O3/c1-4-7-5(2)9-6(3)8-4/h4-6H,1-3H3

InChI key

SQYNKIJPMDEDEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Modified Sanford reaction
  • Synthesis of N-arylbenzoquinaldinium derivatives with antimicrobial activity
  • Hydroformylation of alkenylamines
  • Studies of molecules with inhibitory activity against HIV-1 integrase
  • Irregular Wittig reactions for the formation of α-hydroxyketones

Other Notes

Sales restrictions may apply

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hana Kubová et al.
Epilepsia, 46 Suppl 5, 38-42 (2005-07-01)
To test effects of paraldehyde on behavioral outcome of status epilepticus (SE) in developing rats. Motor SE was induced by LiCl-pilocarpine in rats on postnatal (P) day 12 or 25. Two hours after SE onset, animals were injected with a
Sethu Gunasekaran et al.
Journal of molecular modeling, 14(5), 375-383 (2008-02-27)
The FTIR and Laser-Raman spectra of paraldehyde have been recorded in the regions 4000-400 cm(-1) and 3500-250 cm(-1) respectively. Molecular electronic energy, geometrical structure, harmonic vibrational spectra, infrared intensities and Raman scattering activities have been computed at the HF/6-31G(d,p) and
Will Townend et al.
Emergency medicine journal : EMJ, 19(1), 50-50 (2002-01-05)
A short cut review was carried out to establish whether phenytoin or paraldehyde should be given as the second drug for resistant fits in children. Altogether 41 papers were found using the reported search, of which none answered the question
The role of paraldehyde and lidocaine in the management of status epilepticus.
M Thulasimani et al.
Anaesthesia, 57(1), 99-100 (2002-02-19)
Use of paraldehyde in neonates - an experience.
D Mishra et al.
Indian pediatrics, 38(2), 209-210 (2001-02-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service