Skip to Content
Merck
All Photos(1)

Key Documents

72309

Sigma-Aldrich

Nicotinic acid

≥99.5% (HPLC)

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥99.5% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤0.2% water

ign. residue

≤0.05%

color

colorless to white

mp

234-238 °C
236-239 °C (lit.)

solubility

water: 0.1 g/10 mL, clear, colorless

density

1.473 g/cm3 at 25 °C

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)
rat ... Chrnb2(54239)

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid, also referred as Niacin or vitamin B3, is a water soluble vitamin.[1] It is present in vitamin-enriched bread, liver, meat, fish, poultry, potatoes, green vegetables, peanuts and cereals.[2]

Application

Nicotinic acid has been used as a standard in NAD+ precursor rescue experiments.[3] It has also been used as a vitamin supplement in various culture media such as complete medium (CM)[4] and synthetic medium.[5]

Biochem/physiol Actions

Nicotinic acid or Niacin regulates lipid metabolism by reducing low density lipoprotein (LDL), increasing high-density lipoprotein (HDL) and by inhibiting the progression of atherosclerosis and coronary artery lesions.[2][6] Niacin deficiency leads to pellagra.[2] In addition to vitamin supplement, niacin is also used as a broad spectrum lipid drug.[7]

Other Notes

Coenzyme and Cofactors, vol 2: Pyridine Nucleotide Coenzymes[8][9]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.Dolphin, R.Poulson, O.Avramovic, eds.
Chemical, Biochemical and Medical Aspects, 759-759 (1987)
Nicotinic Acid Regulates Glucose and Lipid Metabolism Through Lipidindependent Pathways
Liu D, et al.
Current Pharmaceutical Biotechnology, 16(1), 3-10 (2015)
Nicotinic acid: the broad-spectrum lipid drug. A 50th anniversary review
Carlson LA
Journal of Internal Medicine, 258(2), 94-114 (2005)
NAMPT-mediated NAD+ biosynthesis in adipocytes regulates adipose tissue function and multi-organ insulin sensitivity in mice
Stromsdorfer KL, et al.
Testing, 16(7), 1851-1860 (2016)
Chemical, Biochemical and Medical Aspects, 776-776 null

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service