Skip to Content
Merck
All Photos(3)

Key Documents

P3075

Sigma-Aldrich

Phenylarsine oxide

≥97%, powder

Synonym(s):

Arzene, Oxophenylarsine, PAO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5AsO
CAS Number:
Molecular Weight:
168.02
Beilstein:
2935227
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic (organic)

Assay

≥97%

form

powder

color

white to off-white

mp

145-148 °C

solubility

DMSO: 50 mg/mL

SMILES string

O=[As]c1ccccc1

InChI

1S/C6H5AsO/c8-7-6-4-2-1-3-5-6/h1-5H

InChI key

BQVCCPGCDUSGOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylarsine oxide (PhAsO4) is a bifunctional SH group chemical. It is hydrophobic in nature.[1]

Application

Phenylarsine oxide has been used to infuse animals with a nigrostriatal transection, to determine whether peroxovanadium (pVa) would synergize with brain-derived neurotrophic factor (BDNF).[2]

Biochem/physiol Actions

Phenylarsine oxide inhibits internalization of cell surface receptors; inhibits tyrosine phosphatases, with no effect on tyrosine kinase. Metabolic poison.
Phenylarsine oxide (PAO) can reduce the motility and sustainability of Setaria cervi.[3] It has the ability to stimulate cyclosporin A-sensitive transition in the presence of EGTA (ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid). It acts as a stimulator of mitochondrial ion fluxes.[1]

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Tyrosine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrosine phosphatase inhibition enhances neurotrophin potency and rescues nigrostriatal neurons in adult rats
Lu X, et al.
Experimental neurology, 178(2), 259-267 (2002)
Inhibition of setaria cervi protein tyrosine phosphatases by phenylarsine oxide: A proteomic and biochemical study
Singh N, et al.
Acta Tropica, 159, 20-28 (2016)
Phenylarsine oxide induces the cyclosporin A-sensitive membrane permeability transition in rat liver mitochondria
Lenartowicz E, et al.
Journal of Bioenergetics and Biomembranes, 23(4), 679-688 (1991)
M Wachtel et al.
Journal of cell science, 112 ( Pt 23), 4347-4356 (1999-11-24)
Regulation of epithelial and endothelial permeability is essential for proper function of compartmentalized organisms, and tyrosine phosphorylation plays an important role in this process. We analyzed the impact of protein tyrosine phosphatase (PTP) inhibition on the structure of endothelial junctional
Sonal Prasad et al.
Communications biology, 3(1), 792-792 (2020-12-29)
The stereocilia of the inner ear sensory cells contain the actin-binding protein radixin, encoded by RDX. Radixin is important for hearing but remains functionally obscure. To determine how radixin influences hearing sensitivity, we used a custom rapid imaging technique to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service