Skip to Content
Merck
All Photos(1)

Documents

P1348

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine

≥97%

Synonym(s):

1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dipalmitoyl, L-α-Phosphatidylethanolamine, dipalmitoyl, L-β,γ-Dipalmitoyl-α-cephalin, DPPE, Dipalmitoyl phosphatidylethanolamine, Hexadecanoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, PE(16:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H74NO8P
CAS Number:
Molecular Weight:
691.96
Beilstein:
1730601
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97%

form

powder

functional group

amine
ester

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCN)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C37H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34,38H2,1-2H3,(H,41,42)/t35-/m1/s1

InChI key

SLKDGVPOSSLUAI-PGUFJCEWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine is used in model membrane .

Biochem/physiol Actions

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine is a phospholipid, a lipid component in cell membrane .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jarungjit Rujiviphat et al.
Journal of molecular biology, 427(16), 2599-2609 (2015-03-19)
Large GTPases of the dynamin superfamily promote membrane fusion and division, processes that are crucial for intracellular trafficking and organellar dynamics. To promote membrane scission, dynamin proteins polymerize, wrap around, and constrict the membrane; however, the mechanism underlying their role
Hanping Wu et al.
Ultrasound in medicine & biology, 45(9), 2502-2514 (2019-06-30)
Our group recently presented a simple strategy using the non-ionic surfactant, Pluronic, as a size control excipient to produce nanobubbles in the 100-nm range, which exhibited stability and echogenicity on par with clinically available microbubbles. The objective of the present
Muhammad S Khan et al.
European biophysics journal : EBJ, 45(8), 843-852 (2016-08-03)
Black lipid membranes (BLMs) have been used for detecting single-channel activities of pore-forming peptides and ion channels. However, the short lifetimes and poor mechanical stability of suspended bilayers limit their applications in high throughput electrophysiological experiments. In this work, we present
Ilaria Visco et al.
Journal of molecular biology, 428(24 Pt A), 4828-4842 (2016-11-04)
Cell polarity arises from a combination of interactions between biological molecules, such as activation, inhibition, and positive or negative feedback between specific polarity units. Activation and inhibition often take place in the form of a membrane binding switch. Lethal giant
Muneyuki Matsuo et al.
Scientific reports, 9(1), 6916-6916 (2019-05-08)
DNA is an essential carrier of sequence-based genetic information for all life today. However, the chemical and physical properties of DNA may also affect the structure and dynamics of a vesicle-based model protocell in which it is encapsulated. To test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service