Skip to Content
Merck
All Photos(2)

Key Documents

850702P

Avanti

12:0 PE

1,2-dilauroyl-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

1,2-didodecanoyl-sn-glycero-3-phosphoethanolamine; DLPE; PE(12:0/12:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H58NO8P
CAS Number:
Molecular Weight:
579.75
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850702P-1g)
pkg of 1 × 200 mg (850702P-200mg)
pkg of 1 × 25 mg (850702P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850702P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O

InChI

1S/C29H58NO8P/c1-3-5-7-9-11-13-15-17-19-21-28(31)35-25-27(26-37-39(33,34)36-24-23-30)38-29(32)22-20-18-16-14-12-10-8-6-4-2/h27H,3-26,30H2,1-2H3,(H,33,34)

InChI key

ZLGYVWRJIZPQMM-UHFFFAOYSA-N

General description

Phosphatidylethanolamine (PE) is one of the widely distributed glycerophospholipid in eukaryotic cells. 1,2-dilauroyl-sn-glycero-3-phosphoethanolamine (12:0 PE/DLPE) possess an ammonium head group (RNH3+) and has alkyl chains made of 11 carbon atoms.

Application

1,2-dilauroyl-sn-glycero-3-phosphoethanolamine (12:0 PE):
  • has been used as a lipid standard in the optimization of the liquid chromatography−mass spectrometry (LC-MS) method conditions
  • may be used as a phospholipid (PL) standard to quantify phosphatidylethanolamine (PE) from plant lipids
  • may be used to prepare anionic liposomes
  • may be used to prepare binary giant unilamellar vesicles (GUVs)

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a vital role in the structure and function of eukaryotic membranes.

Packaging

20 mL Clear Glass Screw Cap Vial (850702P-1g)
5 mL Amber Glass Screw Cap Vial (850702P-200mg)
5 mL Amber Glass Screw Cap Vial (850702P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of DOPC on the permeability of lipid membrane: Detergent solubilization and NMR study
Mady MM
Romanian Journal of Biophysics, 17, 259-267 (2007)
Oligonucleotide delivery with serum into HeLa cells using polycation liposomes
Mady MM
Journal of Biomedical Materials Research. Part B, Applied Biomaterials, 7(1), 69-69 (2007)
K V Damodaran et al.
Biophysical journal, 66(4), 1076-1087 (1994-04-01)
A 250 ps molecular dynamics simulation of the dimyristoylphosphatidylcholine (DMPC)-based lipid bilayer, including explicit water molecules, is reported. The solvent environment of the head groups and other structural properties of the bilayer have been analyzed and compared with experimental results
Takehiro Jimbo et al.
Biophysical journal, 110(7), 1551-1562 (2016-04-14)
We investigate a temperature-driven recursive division of binary giant unilamellar vesicles (GUVs). During the heating step of the heating-cooling cycle, the spherical mother vesicle deforms to a budded limiting shape using up the excess area produced by the chain melting
Phosphatidylethanolamine metabolism in health and disease
International Review of Cell and Molecular Biology, 321, 29-88 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service