Skip to Content
Merck
All Photos(2)

Key Documents

P0121

Sigma-Aldrich

Anti-Purinergic Receptor P2X3 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry: suitable using rat embryo DRG
western blot: 1:200 using rat dorsal root ganglion (DRG) lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The P2X3 (purinergic receptor P2X 3) receptor is expressed in primary afferent neurons. It is a subtype of P2X receptors family of ligand-gated ion channels, activated by extracellular ATP (adenosine triphosphate).

Immunogen

peptide (C)VEKQSTDSGAYSIGH corresponding to residues 383-397 of rat P2rx3.

Application

Anti-Purinergic Receptor P2X3 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

The P2X3 (purinergic receptor P2X 3) plays a major role in the transmission of pain. Inducing P2X3 generates a desensitizing transient response and initiating P2X2/3 creates a relatively persistent response. P2X3 decreases the sympathetic tone in SH rats.

Target description

The P2X3 receptor is highly expressed on nociceptive sensory neurons in dorsal root ganglia (DRG) as a homomer or as a heteromer (P2X3/P2X2). ATP released from damaged cells activates the P2X3 receptor to initiate nociceptive signals. Involvement of ATP in the mechanism of chronic pain contributes to P2X3 receptor as a possible target for the development of pain therapeutics.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bradykinin contributes to sympathetic and pressor responses evoked by activation of skeletal muscle afferents P2X in heart failure.
Xing J and Li J
Cellular Physiology and Biochemistry, 39(6), 2101-2109 (2016)
LncRNA uc. 48+ is involved in diabetic neuropathic pain mediated by the P2X 3 receptor in the dorsal root ganglia.
Wang S, et al.
Purinergic Signaling, 12(1), 139-148 (2016)
P2X3 receptor involvement in endometriosis pain via ERK signaling pathway.
Ding S, et al.
PLoS ONE, 12(9), e0184647-e0184647 (2017)
Purinergic receptors in the carotid body as a new drug target for controlling hypertension.
Pijacka W, et al.
Nature Medicine, 22(10), 1151-1151 (2016)
S Kambiz et al.
Experimental neurology, 267, 87-94 (2015-03-10)
Mirror-image pain is a phenomenon in which unprovoked pain is detected on the uninjured contralateral side after unilateral nerve injury. Although it has been implicated that enhanced production of nerve growth factor (NGF) in the contralateral dorsal root ganglion is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service